Skip to Content
Merck
All Photos(2)

Key Documents

T7782

Sigma-Aldrich

Anti-Mouse IgG (Fab specific)–TRITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

TRITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

direct immunofluorescence: 1:64
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:400

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin G (IgG)antibody is found in four subclasses IgG1, IgG2, IgG3 and IgG4. It has a monomeric structure. IgG is secreted by B cells and is found in blood and extracellular fluids.. Anti-Mouse IgG (Fab specific) is conjugated to tetramethylrhodamine isothiocyanate (TRITC) and then purified by gel filtration to remove free TRITC.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Conjugation of TRITC to Mouse IgG (Fab specific) antibody enables the use in immunofluorescent techniques.

Biochem/physiol Actions

IgG antibody subtype provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.
Immunoglobulin G (IgG) antibody participates in complement fixation and opsonization.

Other Notes

Antibody adsorbed with bovine, equine and human serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background staining with bovine, horse or human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Voldstedlund et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 105(7), 537-545 (1997-07-01)
Most cells express facilitative glucose transporters. Four isoforms (GLUT1-4) transporting D-glucose across the plasma membrane show a specific tissue distribution, which is the basis for tissue-specific patterns in glucose metabolism. GLUT1 is expressed at high levels in tissue barriers such
Hua Jiang et al.
Molecular oncology, 8(2), 297-310 (2014-01-07)
Reprogramming of NK cells with a chimeric antigen receptor (CAR) proved an effective strategy to increase NK cell reactivity and recognition specificity toward tumor cells. To enhance the cytotoxicity of NK cells against CD138-positive multiple myeloma (MM) cells, we generated
Yi-Liang Miao et al.
Environmental and molecular mutagenesis, 50(8), 666-671 (2009-07-14)
Centrosomes are critically important for maintaining meiotic spindle integrity in the meiosis II (MII) stage where oocytes are arrested in most mammalian species before fertilization takes place. In women of advanced ages or during in vitro fertilization (IVF) procedures, aneuploidy
Sh Hejazi et al.
Iranian journal of parasitology, 7(2), 53-60 (2012-10-31)
Cutaneous leishmaniasis is a neglected parasitic disease, which imposes massive human distress and financial costs to the endemic countries. Better understanding of host immune response to the parasite leads to helpful strategies for disease control. Interleukin (IL)-10 and transforming growth
Sh Hejazi et al.
Iranian journal of parasitology, 7(3), 16-23 (2012-10-31)
Cutaneous leishmaniasis is a neglected parasitic disease, which imposes massive human distress and financial costs to the endemic countries. Better understanding of host immune response to the parasite leads to helpful strategies for disease control. Interleukin (IL)-10 and transforming growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service