Skip to Content
Merck
All Photos(3)

Key Documents

T4891

Sigma-Aldrich

Glyceryl tridodecanoate

≥99%

Synonym(s):

1,2,3-Tridodecanoylglycerol, 1,2,3-Trilauroylglycerol, Glycerol trilaurate, Glyceryl trilaurate, Tridodecanoin, Trilaurin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)10COOCH2]2CHOCO(CH2)10CH3
CAS Number:
Molecular Weight:
639.00
Beilstein:
1730452
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

plant

Assay

≥99%

form

powder

mp

46.5 °C (lit.)

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC

InChI

1S/C39H74O6/c1-4-7-10-13-16-19-22-25-28-31-37(40)43-34-36(45-39(42)33-30-27-24-21-18-15-12-9-6-3)35-44-38(41)32-29-26-23-20-17-14-11-8-5-2/h36H,4-35H2,1-3H3

InChI key

VMPHSYLJUKZBJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Development of new lipid-based paclitaxel nanoparticles using sequential simplex optimization.: This article presents the development of lipid-based nanoparticles for drug delivery, utilizing glyceryl tridodecanoate as a component. The optimized nanoparticles show potential for improved delivery and efficacy of paclitaxel, a chemotherapeutic agent (Dong et al., 2009).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Ferstl et al.
Annals of the New York Academy of Sciences, 1189, 62-67 (2010-03-18)
The melting point of triacylglycerides (TAGs) under atmospheric pressure depends on both the fatty acid composition and crystalline structure of the polymorphic state, which are influenced by the temperature treatment history of the TAG. In this contribution, the additional effect
Gianluca Santarossa et al.
FEBS letters, 579(11), 2383-2386 (2005-04-26)
The cold-adapted Pseudomonas fragi lipase (PFL) displays highest activity on short-chain triglyceride substrates and is rapidly inactivated at moderate temperature. Sequence and structure comparison with homologous lipases endowed with different substrate specificity and stability, pointed to three polar residues in
S Iu Zaĭtsev et al.
Bioorganicheskaia khimiia, 26(3), 224-230 (2000-05-19)
A simple method for determining the enzymic hydrolysis parameters of lipid-like substrates and trilaurin assembled in monolayers at the water-air interface was suggested. At a surface pressure of 10 mN/m, the initial rates of lipolysis were found to be proportional
D L Dorset
Biochimica et biophysica acta, 1047(2), 112-120 (1990-11-12)
Binary phase behavior of saturated cholesterol esters with trilaurin or cholesterol, and cholesterol with trilaurin, is studied. The existence of specific molecular interactions is detected by comparing the liquidus curve of the eutectic with ideal theory of freezing point depression
H Heiati et al.
Pharmaceutical research, 13(9), 1406-1410 (1996-09-01)
Solid lipid nanoparticles (SLN) are comprised of a high-melting point triglyceride (TG) core with a phospholipid (PL) coating. This study has investigated the possible formation of multiple PL bilayers on the TG core of SLN's as a function of increasing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service