Skip to Content
Merck
All Photos(1)

Key Documents

T2400

Sigma-Aldrich

Tropomyosin from porcine muscle

lyophilized powder (contains Tris buffer salts)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

Porcine muscle

Quality Level

Assay

≥70% protein basis (biuret)

form

lyophilized powder (contains Tris buffer salts)

technique(s)

electrophoresis: suitable

storage temp.

−20°C

General description

Tropomyosin is a member of the actin filament binding protein family. It is a ubiquitous protein present in animals and fungi. Tropomyosin is made up of two α-helical chains arranged as a coiled-coil. It exists over twenty isoforms, in different eukaryotic cells like skeletal, cardiac, and smooth muscles, brain, platelets, fibroblasts, platelets, and other non-muscle cells.

Application

Tropomyosin from porcine muscle has been used:

  • as a positive control in actin- α-tropomyosin (TPM1) co-sedimentation analysis and assay
  • in analyzing the anti-adhesive properties of fish tropomyosins
  • in far Western blot analysis

Biochem/physiol Actions

Dimeric protein that assumes an α-helical coiled coil conformation. In the absence of Ca2+, inhibits muscle contractility by blocking the myosin binding sites on actin.
Tropomyosin is associated with the actin cytoskeleton and plays a role in stabilizing and regulating the interactions of actin with other actin-binding proteins. It also plays a role in muscle contraction by regulating the interactions of actin with myosin.
Tropomyosin together with troponin, regulates the binding of myosin to actin. Tropomyosin is a dimeric coiled-coil protein forming continuous polymers along the myosin-binding groove of actin. Various tropomyosin isoforms help confer structure and function to actins in the cytoskeleton and striated muscle function. Dimeric protein that assumes an α-helical coiled-coil conformation. In the absence of Ca2+, it inhibits muscle contractility by blocking the myosin binding sites on actin.

Analysis Note

SDS electrophoresis shows two major bands.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel alpha-tropomyosin mutation associates with dilated and non-compaction cardiomyopathy and diminishes actin binding
Judith B A, et al.
Biochimica et Biophysica Acta - Molecular Cell Research, 1833(4), 833-839 (2013)
Vertebrate tropomyosin: distribution, properties and function
Perry S V, et al.
Journal of Muscle Research and Cell Motility, 5?49-5?49 (2001)
A systematic nomenclature for mammalian tropomyosin isoforms
Michael A G, et al.
Journal of Muscle Research and Cell Motility, 147?153-147?153 (2015)
The molecular basis for tropomyosin isoform diversity
James P L M, et al.
Bioessays (1991)
M I Hamilton et al.
Clinical and experimental immunology, 99(3), 404-411 (1995-03-01)
Ulcerative colitis (UC) has a proposed autoimmune pathogenesis. A 40-kD antigen (P40) has been isolated from UC colon, bound to immunoglobulin. Tropomyosin has been reported as the target antigen of a MoAb (7E12H12) raised against P40. We set out to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service