Skip to Content
Merck
All Photos(1)

Key Documents

T1892

Sigma-Aldrich

Thioflavine S

Practical grade, powder

Synonym(s):

Direct Yellow 7

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Colour Index Number:
49010
EC Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

Product Name

Thioflavine S, practical grade

grade

practical grade

form

powder

technique(s)

microbe id | staining: suitable

color

light beige to dark beige, to Dark Brown

solubility

ethanol: water (1:1): 1 mg/mL

λmax

360-380

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

S1c2c(ccc(c2)C)N(C1=C3C=CC(=[N+](C)C)C=C3)C.[Cl-]

InChI

1S/C17H19N2S.ClH/c1-12-5-10-15-16(11-12)20-17(19(15)4)13-6-8-14(9-7-13)18(2)3;/h5-11H,1-4H3;1H/q+1;/p-1

InChI key

JADVWWSKYZXRGX-UHFFFAOYSA-M

General description

Thioflavine S aids in staining and visualizing β-amyloid plaques in Alzheimer′s disease. Thioflavin S binds to fibrillar β-amyloid plaques, and does not diffuse in the plaques. Thioflavin-S staining aids in visualizing smaller plaques throughout the neocortex, hippocampus and thalamic region.

Application

Thioflavine S has been used to stain and visualize β-amyloid structures in the tissue sections.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic modulation of soluble A beta rescues cognitive and synaptic impairment in a mouse model of Alzheimer's disease
Fowler SW, et al.
The Journal of Neuroscience, 34(23), 7871-7885 (2014)
MRI and histological analysis of beta-amyloid plaques in both human Alzheimer's disease and APP/PS1 transgenic mice
Meadowcroft MD, et al.
Journal of Magnetic Resonance Imaging : JMRI, 29(5), 997-1007 (2009)
Disha Shah et al.
PloS one, 8(12), e84241-e84241 (2013-12-21)
Functional connectivity (FC) studies have gained immense popularity in the evaluation of several neurological disorders, such as Alzheimer's disease (AD). AD is a complex disorder, characterised by several pathological features. The problem with FC studies in patients is that it
Attenuation of beta-amyloid deposition and neurotoxicity by chemogenetic modulation of neural activity
Yuan P and Grutzendler J
The Journal of Neuroscience, 36(2), 632-641 (2016)
Exacerbation of cerebral amyloid angiopathy-associated microhemorrhage in amyloid precursor protein transgenic mice by immunotherapy is dependent on antibody recognition of deposited forms of amyloid beta
Racke MM, et al.
The Journal of Neuroscience, 25(3), 629-636 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service