Skip to Content
Merck
All Photos(3)

Key Documents

T0388

Sigma-Aldrich

Tartrazine

Dye content ≥85 %, Powder

Synonym(s):

Acid Yellow 23

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H9N4Na3O9S2
CAS Number:
Molecular Weight:
534.36
Colour Index Number:
19140
Beilstein:
69850
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.47

Product Name

Tartrazine, Dye content ≥85 %

form

powder

composition

Dye content, ≥85%

technique(s)

titration: suitable

color

orange

solubility

H2O: 10 mg/mL, clear, yellow to orange

density

2.12 at 20 °C

εmax

435-500 at 425-429 nm in water

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[Na+].[O-]C(=O)C1=NN(c2ccc(cc2)S([O-])(=O)=O)C(=O)C1N=Nc3ccc(cc3)S([O-])(=O)=O

InChI

1S/C16H12N4O9S2.3Na/c21-15-13(18-17-9-1-5-11(6-2-9)30(24,25)26)14(16(22)23)19-20(15)10-3-7-12(8-4-10)31(27,28)29;;;/h1-8,13H,(H,22,23)(H,24,25,26)(H,27,28,29);;;/q;3*+1/p-3

InChI key

UJMBCXLDXJUMFB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Tartrazine (Cl 19140) also known as Acid yellow 23, is a hydrophilic polyanionic dye. It is a sulfonic dye, which can be synthesized by the reaction of sulfanilic acid with 3-carboxy-l-(4-sulfophenyl)-5-pyrazolone.

Application

Tartrazine may be used as a reference standard in the determination of tartrazine in food samples using ion-pair liquid chromatography coupled with photodiode-array and electrospray mass spectrometry detector.
The application of Tartrazine is seen in the phloxine-tartrazine procedure for cellular inclusions such as viral aggregates and eosinophil granules. It may be used as a substitute for orange G in a variant of Mallory′s trichrome, and in a methyl violet-tartrazine mixture to stain cryptosporidia in faecal smears and mucosal scrapings.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel C Corbett et al.
Science advances, 6(40) (2020-10-02)
Spatial patterns of gene expression in living organisms orchestrate cell decisions in development, homeostasis, and disease. However, most methods for reconstructing gene patterning in 3D cell culture and artificial tissues are restricted by patterning depth and scale. We introduce a
Jin Yee Gan et al.
Membranes, 10(8) (2020-08-07)
This study produced a novel polysulfone (PSF) membrane for dye removal using lemon-derived carbon quantum dots-grafted silver nanoparticles (Ag/CQDs) as membrane nanofiller. The preparation of CQDs was completed by undergoing hydrothermal treatment to carbonize the pulp-free lemon juice into CQD
Simultaneous analysis of tartrazine and its intermediates by reversed phase liquid chromatography.
Wittmer PD, et al.
Analytical Chemistry, 47(8), 1422-1423 (1975)
Stella A Ordoudi et al.
Food chemistry, 267, 410-419 (2018-06-24)
The stepwise approach takes advantage of simple, versatile, low-cost screening tools that can be applied to several posts of the saffron trade chain to specifically detect adulteration with carminic acid (CA). This natural dye is of insect origin and should
Helen Waechter et al.
Analytical chemistry, 81(21), 9048-9054 (2009-10-10)
Analytical UV absorption detection for microfluidic devices, capillary electrophoresis, and even high-performance liquid chromatography is hampered by the small detection volumes, short absorption paths, and the need to sample at a high rate with a stable background and low noise.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service