SRP6516
Albumin from human plasma
≥95% (SDS-PAGE)
Synonym(s):
Albumin protein, human Albumin, human Albumin protein, human plasma, human plasma Albumin proteinHSA, native Albumin, native Albumin protein
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
General description
Human albumin is encoded by ALB gene that is mapped to human chromosome 4q. The gene is developmentally regulated and is synthesized by the liver. The encoded protein has a molar mass of 65,000 Da and consists of a single polypeptide chain of 585 amino acids.
Application
Albumin has been used in ion exchange chromatography to compare the albumin isolated from whole human plasma. It has been used to bind to phage clones in phage binding assay.
Biochem/physiol Actions
Human albumin is a globular unglycosylated serum protein that makes up for more than half of the total plasma or serum proteins. It functions in the maintenance of COP (cellular oncotic pressure). It binds to hydrophobic organic anions such as, bilirubin, long-chain fatty acids and haematin and some cations such as magnesium and calcium. It also serves as a secondary or tertiary carrier for steroids, vitamin D and thyroxine. It is involved in the catabolism of prostaglandins and metabolism of certain endogenous substances, such as lipids, and eicosanoids. It acts as a plasma buffer and scavenges oxygen free radicals, thus reducing the pathological effects of certain inflammatory diseases. Mutation in the gene coding for albumin leads to familial dysalbuminaemic hyperthyroidism.
Physical form
Lyophilized as a salt free solid.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1A
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Isolation of albumin from whole human plasma and fractionation of albumin-depleted plasma.
The Biochemical Journal, 157, 301-306 (1976)
Molecular structure of the human albumin gene is revealed by nucleotide sequence within q11-22 of chromosome 4.
The Journal of Biological Chemistry, 261, 6747-6757 (1986)
Cardiovascular research, 117(3), 820-835 (2020-04-08)
Calcific aortic valve disease (CAVD) is the most common heart valve disease in the Western world. It has been reported that zinc is accumulated in calcified human aortic valves. However, whether zinc directly regulates CAVD is yet to be elucidated.
The role of albumin in critical illness.
British Journal of Anaesthesia, 85, 599-610 (2000)
Albumin binding as a general strategy for improving the pharmacokinetics of proteins.
The Journal of Biological Chemistry, 277, 35035-35043 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service