Skip to Content
Merck
All Photos(1)

Documents

SRP6006

Sigma-Aldrich

IL-1RA murine

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

ICIL-1RA, IL-1ra3, IL1 inhibitor, IL1F3, IRAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized

mol wt

17.4 kDa

packaging

pkg of 50 μg

technique(s)

activity assay: suitable

impurities

Endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... IL-1RA(16181)

General description

Research area: IMMUNO AND CKS

Interleukin-1 receptor antagonist (IL-1 RA) is a member of the interleukin 1 cytokine family. Four alternatively spliced transcript variants encoding distinct isoforms have been reported. IL-1RA mouse recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 153 amino acids and having a molecular mass of 17.4kDa. The IL1RA is purified by proprietary chromatographic techniques. This gene and five other closely related cytokine genes form a gene cluster spanning approximately 400 kb on chromosome 2.
Interleukin-1 RA is a member of the interleukin 1 cytokine family. This protein inhibits the activities of interleukin 1-alpha (IL1A) and interleukin 1-beta (IL1B), and modulates a variety of interleukin 1 related immune and inflammatory responses. This gene and five other closely related cytokine genes form a gene cluster spanning approximately 400 kb on chromosome 2. A polymorphism of this gene is reported to be associated with increased risk of osteoporotic fractures and gastric cancer. Four alternatively spliced transcript variants encoding distinct isoforms have been reported. IL1 ra Mouse Recombinant produced in E.Coli is a single, non-glycosylated polypeptide chain containing 153 amino acids and having a molecular mass of 17.4kDa. The IL1RA is purified by proprietary chromatographic techniques.

Application

IL-1RA murine has been used as an antagonist to the interleukin-1 receptor in fibroblasts.

Biochem/physiol Actions

Interleukin-1 receptor antagonist (IL-1Ra) protein mediates the inhibition of interleukin 1-α (IL1A) and interleukin 1-β (IL1B) activities. It is also involved in the modulation of IL-1 related immune and inflammatory responses. A polymorphism in the IL-1Ra gene is reported to be associated with an increased risk of osteoporotic fractures and gastric cancer. IL-1Ra is effective in treating bronchopulmonary dysplasia in murine models.This protein inhibits the activities of interleukin 1-alpha (IL1A) and interleukin 1-beta (IL1B) and modulates a variety of interleukin 1 related immune and inflammatory responses.

Physical form

Lyophilized from H?O containing NaHCO?

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile ddH?O to a concentration ? 100 μg/ml. This solution can then be diluted into other aqueous buffers.

Other Notes

The IL1RA is purified by proprietary chromatographic techniques.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W P Arend et al.
Annual review of immunology, 16, 27-55 (1998-05-23)
The interleukin-1 receptor antagonist (IL-1Ra) is a member of the IL-1 family that binds to IL-1 receptors but does not induce any intracellular response. Two structural variants of IL-1Ra have previously been described: a 17-kDa form that is secreted from
Christine B Bui et al.
Frontiers in immunology, 10, 1480-1480 (2019-07-30)
Pulmonary hypertension secondary to bronchopulmonary dysplasia (BPD-PH) represents a major complication of BPD in extremely preterm infants for which there are currently no safe and effective interventions. The abundance of interleukin-1 (IL-1) is strongly correlated with the severity and long-term
The role of interleukin-1 polymorphisms in the pathogenesis of gastric cancer.
E M El-Omar et al.
Nature, 412(6842), 99-99 (2002-01-26)
Athanasios Didangelos et al.
Scientific reports, 6, 21607-21607 (2016-02-24)
Spinal cord injury is characterized by acute cellular and axonal damage followed by aggressive inflammation and pathological tissue remodelling. The biological mediators underlying these processes are still largely unknown. Here we apply an innovative proteomics approach targeting the enriched extracellular
Hui Zhao et al.
Journal of neuroinflammation, 8, 159-159 (2011-11-15)
Neuroimmune modulation following traumatic stress is accompanied by cortical upregulation of c-Src expression, but the mechanistic details of the potential regulatory link between c-Src expression and immunosuppression have not been established. We used a combination of techniques to measure temporal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service