Skip to Content
Merck
All Photos(1)

Key Documents

SRP3267

Sigma-Aldrich

NT-3 human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

HGNF, NGF-2, Nerve growth factor-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

20-50 ng/mL ED50

mol wt

27.2 kDa

packaging

pkg of 10 μg

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NTF3(4908)

General description

NTF3 (neurotrophin 3) is a neurotrophic factor belonging to the NGF (nerve growth factor) family. This protein is composed of 257 amino acids, with a prepro region of 138 amino acids and the mature sequence of 119 residues.

Biochem/physiol Actions

NT3 functions on two structurally unrelated receptor types- the p75 neurotrophin receptor (p75NTR) and the receptor tyrosine kinase (TRK). It promotes the metastatic invasion of brain by breast cancer cells, by promoting the EMT (epithelial-mesenchymal transition) transition of breast cancer cells and suppressing the immune response in brain. This protein is responsible for the indction and survival of neurite outgrowth from a subset of neural crest (NC) and placode-derived neurons.
Recombinant human NT-3 is a noncovalently linked homodimer, of two 13.6kDa polypeptide monomers (240 total amino acid residues).

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neurotrophin-3 modulates breast cancer cells and the microenvironment to promote the growth of breast cancer brain metastasis.
Oncogene, 32(35), 4064-4077 (2013)
Neurotrophin 3 is a mitogen for cultured neural crest cells.
Kalcheim C et al
Proceedings of the National Academy of Sciences of the USA, 89(5), 1661-1665 (1992)
Cloning and expression of a cDNA encoding a novel human neurotrophic factor.
Kaisho Y et al
Febs Letters, 266(1-2), 187-191 (1990)
P C Maisonpierre et al.
Genomics, 10(3), 558-568 (1991-07-01)
The development and maintenance of the vertebrate nervous system depends upon neuronal survival proteins known as neurotrophic factors. Nerve growth factor (NGF) remains the best characterized neurotrophic molecule. Brain-derived neurotrophic factor (BDNF) and neurotrophin-3 (NT-3) are two recently cloned neurotrophic
Ke Zhang et al.
Stem cell reviews, 10(4), 612-625 (2014-05-03)
In the past decades, mesenchymal stem cells (MSCs) as a promising cell candidate have received the most attention in the treatment of spinal cord injury (SCI). However, due to the low survival rate and low neural differentiation rate, the grafted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service