Skip to Content
Merck
All Photos(2)

Key Documents

SML3846

Sigma-Aldrich

Motixafortide trifluoroacetate

≥95% (HPLC)

Synonym(s):

TF 14016 trifluoroacetate, B 4F-benzoyl-TN14003 trifluoroacetate, BKT 140 trifluoroacetate, BKT140 trifluoroacetate, N2-(4-Fluorobenzoyl)-L-arginyl-L-arginyl-3-(2-naphthalenyl)-L-alanyl-L-cysteinyl-L-tyrosyl-N5-(aminocarbonyl)-L-ornithyl-L-lysyl-D-lysyl-L-prolyl-L-tyrosyl-L-arginyl-N5-(aminocarbonyl)-L-ornithyl-L-cysteinyl- L-argininamide cyclic (4→13)-disulfide trifluoroacetate, TF14016 trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C97H144FN33O19S2 · xC2HF3O2
Molecular Weight:
2159.52 (free base basis)
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

SMILES string

O=C(NCCCC1NC(C(CCCNC(N)=N)NC(C(CC2=CC=C(C=C2)O)NC(C3N(CCC3)C(C(CCCCN)NC(C(CCCCN)NC(C(CCCNC(N)=O)NC(C(CC4=CC=C(C=C4)O)NC(C(CSSCC(C(NC(CCCNC(N)=N)C(N)=O)=O)NC1=O)NC(C(CC5=CC6=C(C=CC=C6)C=C5)NC(C(CCCNC(N)=N)NC(C(CCCNC(N)=N)NC(C7=CC=C(C=C7)F)=O)=O)=O)=O)=O)=O

Biochem/physiol Actions

Motixafortide (BKT140, BL-8040) is an orally available, highly selective inverse agonist of CXCR4 (CXC Chemokine Receptor 4). Motixafortide is a potent stem cell mobiliser and anticancer agent. BKT140 potently inhibits CXCL12 induced migration/invasion of small-cell lung cancer cells.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The CXCR4 inhibitor BL-8040 induces the apoptosis of AML blasts by downregulating ERK, BCL-2, MCL-1 and cyclin-D1 via altered miR-15a/16-1 expression
Leukemia, 31(11), 2336-2346 (2017)
Enhanced unique pattern of hematopoietic cell mobilization induced by the CXCR4 antagonist 4F-benzoyl-TN14003
Stem Cells, 25(9), 2158-2166 (2007)
Targeting the CD20 and CXCR4 pathways in non-hodgkin lymphoma with rituximab and high-affinity CXCR4 antagonist BKT140
Clinical Cancer Research, 19(13), 3495-3507 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service