Skip to Content
Merck
All Photos(1)

Key Documents

SML3237

Sigma-Aldrich

G-5555

≥98% (HPLC)

Synonym(s):

8-(((2r,5r)-5-Amino-1,3-dioxan-2-yl)methyl)-6-(2-chloro-4-(6-methylpyridin-2-yl)phenyl)-2-(methylamino)pyrido[2,3-d]pyrimidin-7(8H)-one, 8-[(trans -5-Amino-1,3-dioxan-2-yl)methyl]-6-[2-chloro-4-(6-methyl-2-pyridinyl)phenyl]-2-(methylamino)pyrido[2,3-d ]pyrimidin-7(8H )-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H25ClN6O3
CAS Number:
Molecular Weight:
492.96
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/L, clear

storage temp.

−20°C

General description

G-5555 is a selective p21-activated kinase (PAK1) inhibitor. In addition to this it also selectively inhibits eight kinases such as PAK2, PAK3, KHS1, lymphocyte-specific protein tyrosine kinase (LcK), Mammalian Ste20-like kinase 3 (MST3), Mammalian Ste20-like kinase 4 (MST4), salt-inducible kinase 2(SIK2), and Yeast Sps1/Ste20-related Kinase 1 (YSK1). G-5555 exhibits high stability and low human ether-à-go-go related gene (hERG) activity than other PAK inhibitors including FRAX1036.

Biochem/physiol Actions

ATP site-targeting inhibitor against SIK1/2/3, MST3/4, PAK1/2, YANK1/3, MAP2K5 and MAP4K5. Reduces cellular MEK pS298 in cultures and in mice in vivo.
G-5555 is an ATP site-targeting inhibitor against salt-inducible kinases, MST3/4, p21-activated kinase 1/2, YANK1/3, MAP2K5, MAP4K5 (% inhibition at 100 nM = 98.6/93.5/99.8 (SIK1/2/3), 85/99.5 (MST3/4), 95.6/87 (PAK1/2), 94.7/96.5 (YANK1/3); 87/MAP2K5, 83/MAP4K5) with good selectivity over other kinases. G-5555 downregulates MEK pS298 in EBC1 cells in cultures (IC50 = 69 nM) and in H292 NSCLC tumors in mice in vivo (25 mg/kg p.o.) with good oral availability (mice F = 80%, monkeys F = 72%).
G-5555 is an ATP site-targeting inhibitor against salt-inducible kinases, MST3/4, p21-activated kinase 1/2, YANK1/3, MAP2K5, MAP4K5 (% inhibition at 100 nM = 98.6/93.5/99.8 (SIK1/2/3), 85/99.5 (MST3/4), 95.6/87 (PAK1/2), 94.7/96.5 (YANK1/3); 87/MAP2K5, 83/MAP4K5) with good selectivity over other kinases. G-5555 downregulates MEK pS298 in EBC1 cells in cultures (IC50 = 69 nM) and in H292 NSCLC tumors in mice in vivo (25 mg/kg p.o.) with good oral availability (mice F = 80%, monkeys F = 72%). PAK1 plays a crucial role in cancer progression, inhibiting this protein has the potential to reduce cancer development. Therefore, G-5555 may be a promising candidate for oncology research.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chudi O Ndubaku et al.
ACS medicinal chemistry letters, 6(12), 1241-1246 (2015-12-30)
Signaling pathways intersecting with the p21-activated kinases (PAKs) play important roles in tumorigenesis and cancer progression. By recognizing that the limitations of FRAX1036 (1) were chiefly associated with the highly basic amine it contained, we devised a mitigation strategy to
Hai-Feng Qiao et al.
Journal of reproductive immunology, 140, 103129-103129 (2020-04-26)
The present study was meant for the discovery of the underlying functions of miR-485-5p in ovarian cancer concerning cisplatin resistance in vitro. RT-qPCR assessed the miR-485-5p expression in ovarian cancer cell lines, normal cells and cisplatin-resistant Cell line OVCA433-CR. After
Christina M Knippler et al.
Endocrine-related cancer, 26(8), 699-712 (2019-05-31)
The number of individuals who succumb to thyroid cancer has been increasing and those who are refractory to standard care have limited therapeutic options, highlighting the importance of developing new treatments for patients with aggressive forms of the disease. Mutational
Hai-Feng Qiao et al.
Journal of reproductive immunology, 140, 103129-103129 (2020-04-26)
The present study was meant for the discovery of the underlying functions of miR-485-5p in ovarian cancer concerning cisplatin resistance in vitro. RT-qPCR assessed the miR-485-5p expression in ovarian cancer cell lines, normal cells and cisplatin-resistant Cell line OVCA433-CR. After
Roberta Tesch et al.
Journal of medicinal chemistry, 64(12), 8142-8160 (2021-06-05)
Salt-inducible kinases (SIKs) are key metabolic regulators. The imbalance in SIK function is associated with the development of diverse cancers, including breast, gastric, and ovarian cancers. Chemical tools to clarify the roles of SIK in different diseases are, however, sparse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service