Skip to Content
Merck
All Photos(1)

Key Documents

SML2928

Sigma-Aldrich

Lumiracoxib

≥98% (HPLC)

Synonym(s):

2-[(2-Chloro-6-fluorophenyl)amino]-5-methyl-benzeneacetic acid, 5-Methyl-2-(2′-chloro-6′-fluoroanilino)phenylacetic acid, CGS 35189, CGS-35189, CGS35189, COX 189, COX-189, COX189, LMX, Non-steroidal anti-inflammatory drug

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13ClFNO2
CAS Number:
Molecular Weight:
293.72
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=CC(F)=C1NC2=CC=C(C)C=C2CC(O)=O

InChI

1S/C15H13ClFNO2/c1-9-5-6-13(10(7-9)8-14(19)20)18-15-11(16)3-2-4-12(15)17/h2-7,18H,8H2,1H3,(H,19,20)

InChI key

KHPKQFYUPIUARC-UHFFFAOYSA-N

Biochem/physiol Actions

Lumiracoxib (COX189) is an orally active, potent and selective cyclooxygenase-2 inhibitor (Ki = 60 nM/COX-2 vs. 3.2 μM/COX-1) that inhibits COX-2-mediated PGE2 production in human whole blood (IC50 = 130 nM; stimulation = 50 μM A23187), but not COX-1-dependent TxB2 production (IC50 = 67 μM; stimulation = 10 μg/mL LPS). Lumiracoxib shows in vivo anti-inflammatory efficacy against carrageenan-induced paw oedema (ED30 = 0.35 mg/kg p.o.), CFA-induced hyperalgesia (ED30 = 5.1 mg/kg p.o.), as well as adjuvant-induced arthritis (ED50 = 3 mg/kg/day p.o.) in rats in vivo.
Orally active, potent and selective cyclooxygenase-2 (COX-2) inhibitor with anti-inflammatory efficacy in vivo.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alyson Fox et al.
Pain, 107(1-2), 33-40 (2004-01-13)
Chronic pain resulting from metastatic bone cancer remains poorly understood and resistant to treatment. Here we have examined the effect of the novel COX-2 enzyme inhibitor lumiracoxib in a model of bone cancer pain in the rat. Lumiracoxib was administered
Weijie Jiao et al.
Drug testing and analysis, 12(6), 827-835 (2020-02-12)
Lumiracoxib is a selective cyclooxygenase-2 inhibitor, which has been reported to cause rare but severe liver injury. Considering that lumiracoxib has a carboxylic group in the molecule, glucuronidation to form acylglucuronide would be one of the possible mechanisms of lumiracoxib-induced
S S Ahmed et al.
Toxicology and applied pharmacology, 369, 39-48 (2019-02-16)
The occurrence of drug hypersensitivity reactions (DHRs) following administration of low molecular weight (LMW) drugs is an important health concern. However, in vivo animal models which could be used as tools for the prediction of DHRs are lacking. As a
Ronald Esser et al.
British journal of pharmacology, 144(4), 538-550 (2005-01-19)
1. This manuscript presents the preclinical profile of lumiracoxib, a novel cyclooxygenase-2 (COX-2) selective inhibitor. 2. Lumiracoxib inhibited purified COX-1 and COX-2 with K(i) values of 3 and 0.06 microM, respectively. In cellular assays, lumiracoxib had an IC(50) of 0.14
Amanda J Morgan et al.
ACS chemical neuroscience, 9(7), 1552-1559 (2018-05-04)
Cyclooxygenase-2 (COX-2) catalyzes the formation of prostaglandins, which are involved in immune regulation, vascular function, and synaptic signaling. COX-2 also inactivates the endogenous cannabinoid (eCB) 2-arachidonoylglycerol (2-AG) via oxygenation of its arachidonic acid backbone to form a variety of prostaglandin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service