Skip to Content
Merck
All Photos(1)

Key Documents

SML2850

Sigma-Aldrich

Cediranib

≥98% (HPLC)

Synonym(s):

4-(4-Fluoro-2-methylindol-5-yloxy)-6-methoxy-7-[3-(pyrrolidin-1-yl)propoxy]quinazoline, 4-[(4-Fluoro-2-methyl-1H-indol-5-yl)oxy]-6-methoxy-7-[3-(1-pyrrolidinyl)propoxy]quinazoline, 4-[(4-Fluoro-2-methyl-1H-indol-5-yl)oxy]-6-methoxy-7-[3-(pyrrolidin-1-yl)propoxy]quinazoline, AZD 2171, AZD2171

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H27FN4O3
CAS Number:
Molecular Weight:
450.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c2c([nH]c(c2)C)ccc1Oc3ncnc4c3cc(c(c4)OCCCN5CCCC5)OC

InChI

1S/C25H27FN4O3/c1-16-12-17-19(29-16)6-7-21(24(17)26)33-25-18-13-22(31-2)23(14-20(18)27-15-28-25)32-11-5-10-30-8-3-4-9-30/h6-7,12-15,29H,3-5,8-11H2,1-2H3

InChI key

XXJWYDDUDKYVKI-UHFFFAOYSA-N

Biochem/physiol Actions

Cediranib (AZD2171) is an orally active anticancer agent and a highly potent ATP-competitive receptor tyrosine kinase (RTK) inhibitor against VEGFR (IC50 = 5 nM/Flt-1 (VEGFR1), <1 nM/KDR (VEGFR2), ≤3 nM/Flt-4 (VEGFR3), c-Kit (IC50 = 2 nM), PDGFR1/2 (IC50 = 5/36 nM), and FGFR1 (IC50 = 26 nM). Cediranib exhibits reduced activity against CSF-1R, Src, Abl (IC50 = 110, 130, 260 nM, respectively) and little or no potency toward Flt-3, EGFR, ErbB2 (Her-2/neu), CDK2/4, Aurora A/B, and MEK.
Orally active anticancer agent, highly potent ATP-competitive tyrosine kinase inhibitor against VEGFR1/2/3 (Flt-1/KDR/Flt-4), c-Kit, PDGFR1/2 & FGFR1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathy D Miller et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(1), 281-288 (2006-01-07)
This pilot study combined physiologic imaging, microcomputed tomography, and histologic tumor evaluation with a xenograft model of breast cancer to identify surrogates likely to correlate with response to AZD2171, an inhibitor of the vascular endothelial growth factor (VEGF) receptor tyrosine
A L R Bordinhão et al.
Oncology reports, 36(6), 3197-3206 (2016-10-18)
Cediranib, a pan-tyrosine kinase inhibitor is showing promising results for the treatment of several solid tumours. In breast cancer, its effects remain unclear, and there are no predictive biomarkers. Several studies have examined the expression profiles of microRNAs (miRNAs) in response
Ji Yeong Kim et al.
Anticancer research, 39(7), 3785-3793 (2019-07-03)
This study investigated drugs able to sensitize P-glycoprotein (P-gp)-overexpressing resistant KBV20C cancer cells to vincristine or eribulin treatment and assessed their associated mechanisms of action. Eight tyrosine kinase inhibitors (lapatinib, gefitinib, imatinib, erlotinib, nilotinib, pazopanib, cediranib, and vandetanib) and one
Stephen R Wedge et al.
Cancer research, 65(10), 4389-4400 (2005-05-19)
Inhibition of vascular endothelial growth factor-A (VEGF) signaling is a promising therapeutic approach that aims to stabilize the progression of solid malignancies by abrogating tumor-induced angiogenesis. This may be accomplished by inhibiting the kinase activity of VEGF receptor-2 (KDR), which
Carolyn Cao et al.
Cancer research, 66(23), 11409-11415 (2006-12-06)
The vascular endothelial growth factor receptor (VEGFR) tyrosine kinases are being explored as targets for antiangiogenic cancer therapy. Radiotherapy also inhibits tumor growth and affects vasculature. We investigated the combination of the potent VEGFR tyrosine kinase inhibitor AZD2171 and ionizing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service