Skip to Content
Merck
All Photos(1)

Key Documents

SML2758

Sigma-Aldrich

Dapagliflozin propanediol monohydrate

≥98% (HPLC)

Synonym(s):

(1S)-1,5-Anhydro-1-C-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-D-glucitol (S)-propane-1,2-diol (1:1) monohydrate, (2S,3R,4R,5S,6R)-2-(3-(4-Ethoxybenzyl)-4-chlorophenyl)-6-hydroxymethyl-tetrahydro-2H-pyran-3,4,5-triol propanediol monohydrate, BMS 512148 propanediol hydrate, BMS-512148 propanediol hydrate, Dapagliflozin (S)-propylene glycol monohydrate, Dapagliflozin propanediol hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25ClO6·C3H8O2·H2O
CAS Number:
Molecular Weight:
502.98
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +17 to +25, c = 1.0 in chloroform-d

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

Clc1c(cc(cc1)[C@@H]3O[C@@H]([C@H]([C@@H]([C@H]3O)O)O)CO)Cc2ccc(cc2)OCC.O[C@H](CO)C.O

InChI

1S/C21H25ClO6.C3H8O2.H2O/c1-2-27-15-6-3-12(4-7-15)9-14-10-13(5-8-16(14)22)21-20(26)19(25)18(24)17(11-23)28-21;1-3(5)2-4;/h3-8,10,17-21,23-26H,2,9,11H2,1H3;3-5H,2H2,1H3;1H2/t17-,18-,19+,20-,21+;3-;/m10./s1

InChI key

GOADIQFWSVMMRJ-UPGAGZFNSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dapagliflozin (BMS-512148) is an orally active, potent and selective renal sodium-dependent glucose cotransporter 2 (SGLT2; SLC5A2) inhibitor (human/rat SGLT2 IC50 = 1.1/3 nM vs. human/rat SGLT1 IC50 = 1.39/0.6 μM; R-methyl-D-glucopyranoside (AMG) uptake assay with respective CHO transfectants). Dapagliflozin reduces blood glucose levels (by 55% in streptozotocin (STZ)-induced hyperglycemic rats 5 hrs post 0.1 mg/kg p.o.) by preventing the renal glucose reabsorption process and promoting glucose excretion in urine with good pharmacokinetic properties and oral availability (84% in rats and 83% in dogs, respectively, post 1 mg/kg and 6.6 mg/kg p.o.) in vivo.
Orally active, potent and selective renal sodium-dependent glucose cotransporter 2 (SGLT2; SLC5A2) inhibitor with blood sugar-lowering efficacy in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Meng et al.
Journal of medicinal chemistry, 51(5), 1145-1149 (2008-02-12)
The C-aryl glucoside 6 (dapagliflozin) was identified as a potent and selective hSGLT2 inhibitor which reduced blood glucose levels in a dose-dependent manner by as much as 55% in hyperglycemic streptozotocin (STZ) rats. These findings, combined with a favorable ADME
Dapagliflozin - a breakthrough in the search for drugs to treat HFrEF.
Karina Huynh
Nature reviews. Cardiology, 16(12), 700-700 (2019-10-04)
Hiroki Yoshioka et al.
Obesity research & clinical practice, 13(5), 505-510 (2019-08-31)
Chronopharmacology is the study of the varying responses of drugs to changes in biological timing and endogenous periodicities. The selective sodium-glucose cotransporter 2 inhibitor, dapagliflozin, is a globally prescribed antihyperglycemic drug. Although dapagliflozin is usually administered once a day, the
Ryo Shibusawa et al.
Scientific reports, 9(1), 9887-9887 (2019-07-10)
The new type 2 diabetes drug, dapagliflozin, reduces blood glucose levels and body weight by inhibiting sodium glucose transporter 2 (SGLT2) in proximal tubular cells. SGLT2 inhibitors might modulate glucose influx into renal tubular cells, thereby regulating the metabolic conditions
Kazuno Omori et al.
Metabolism: clinical and experimental, 98, 27-36 (2019-06-17)
To explore the beneficial effects of dapagliflozin and/or insulin glargine on the pancreatic beta cell mass and hepatic steatosis in db/db mice. Six-week-old db/db mice were assigned to one of four groups: untreated (Placebo), treated with dapagliflozin (Dapa), treated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service