Skip to Content
Merck
All Photos(1)

Key Documents

SML2755

Sigma-Aldrich

Netupitant

≥98% (HPLC)

Synonym(s):

2-(3,5-bis(Trifluoromethyl)phenyl)-N,2-dimethyl-N-(6-(4-methylpiperazin-1-yl)-4-o-tolylpyridin-3-yl)propanamide, 2-[3,5-bis(Trifluoromethyl)phenyl]-N-[6-(4-methylpiperazin-1-yl)-4-o-tolylpyridin-3-yl]-N-methylisobutyramide, AGE 94200, AGE-94200, AGE94200, CID 6451149, CID-6451149, CID6451149, N,a,a-Trimethyl-N-[4-(2-methylphenyl)-6-(4-methyl-1-piperazinyl)-3-pyridinyl]-3,5-bis(trifluoromethyl)benzeneacetamide, Ro 67-31898, Ro 67-31898/000

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H32F6N4O
CAS Number:
Molecular Weight:
578.59
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(cc(c1)C(C)(C)C(=O)N(C)c2cnc(cc2c4c(cccc4)C)N3CCN(CC3)C)C(F)(F)F

InChI

1S/C30H32F6N4O/c1-19-8-6-7-9-23(19)24-17-26(40-12-10-38(4)11-13-40)37-18-25(24)39(5)27(41)28(2,3)20-14-21(29(31,32)33)16-22(15-20)30(34,35)36/h6-9,14-18H,10-13H2,1-5H3

InChI key

WAXQNWCZJDTGBU-UHFFFAOYSA-N

Biochem/physiol Actions

Brain-penetrant, orally active, potent and selective neurokinin-1 (NK1) receptor (NK1R; Tachykinin receptor 1) antagonist with antiemetic efficacy in vivo.
Netupitant is a brain-penetrant, orally active, potent and selective neurokinin-1 (NK1) receptor (NK1R; Tachykinin receptor 1) antagonist (Ca+2 mobilization IC50 = 11.2 nM/hNK1 vs. >1 μM/rNK1 & hNK2/3; hNK1/2/3 Ki = 0.95 nM/1.6 μM/1.6 μM) that effectively blocks agonist GR73632-induced foot tapping in gerbils (ID50 = 0.5 mg/kg po 2h prior to 3 pmol/5 μL GR73632/gerbil icv). In addition to antiemetic efficacy against substance P-associated vomiting reflexes, netupitant is also reported to enhance the analgesic effects of electroacupuncture (EA) among humanized mice with sickle cell disease in response to pain in vivo (10 mg/kg/day, i.p.).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatrice Girard et al.
Experimental neurology, 285(Pt B), 110-125 (2016-10-25)
The mechanisms underlying the postnatal maturation of micturition from a somatovesical to a vesicovesical reflex are not known but may involve neuropeptides in the lower urinary tract. A transgenic mouse model with chronic urothelial overexpression (OE) of NGF exhibited increased
Ying Wang et al.
Journal of pain research, 12, 2419-2426 (2019-08-27)
Purpose: Chronic pain is a major comorbidity of sickle cell disease (SCD). Acupuncture, a non-opioid and non-addictive therapy to treat pain, was found to reduce pain in the majority (80%) of SCD patients in an earlier retrospective review. We observed
John A Rudd et al.
Frontiers in pharmacology, 7, 263-263 (2016-09-16)
Chemotherapy-induced acute and delayed emesis involves the activation of multiple pathways, with 5-hydroxytryptamine (5-HT; serotonin) playing a major role in the initial response. Substance P tachykinin NK1 receptor antagonists can reduce emesis induced by disparate emetic challenges and therefore have
Ajit G Thomas et al.
Experimental brain research, 232(8), 2637-2644 (2014-06-28)
Current therapy for chemotherapy-induced nausea and vomiting includes the use of both 5-HT3 and NK1 receptor antagonists. Acute emesis has largely been alleviated with the use of 5-HT3 receptor antagonists, while an improvement in preventing delayed emesis has been achieved
Marigo Stathis et al.
European journal of pharmacology, 689(1-3), 25-30 (2012-06-12)
Netupitant is a potent and selective NK(1) receptor antagonist under development in combination with a fixed dose of palonosetron for the prevention of chemotherapy induced nausea and vomiting. Palonosetron is a 5-HT(3) receptor antagonist approved for both the prevention of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service