Skip to Content
Merck
All Photos(1)

Key Documents

SML2333

Sigma-Aldrich

VU0463271

≥98% (HPLC)

Synonym(s):

N-Cyclopropyl-N-(4-methyl-2-thiazolyl)-2-[(6-phenyl-3-pyridazinyl)thio]-acetamide, N-Cyclopropyl-N-(4-methyl-2-thiazolyl)-2-[(6-phenyl-3-pyridazinyl)thio]acetamide, N-Cyclopropyl-N-(4-methylthiazol-2-yl)-2-(6-phenylpyridazin-3-ylthio)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N4OS2
CAS Number:
Molecular Weight:
382.50
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

off-white to blue-gray

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c(nc(c1)C)N(C4CC4)C(=O)CSc2nnc(cc2)c3ccccc3

InChI

1S/C19H18N4OS2/c1-13-11-26-19(20-13)23(15-7-8-15)18(24)12-25-17-10-9-16(21-22-17)14-5-3-2-4-6-14/h2-6,9-11,15H,7-8,12H2,1H3

InChI key

DPONSKCACOZTGN-UHFFFAOYSA-N

Biochem/physiol Actions

VU0463271 is a potent and selective potassium-chloride cotransporter 2 (KCC2) inhibitor that displays greatly enhanced potency than its structural analogs VU0255011 (ML077) & VU0240511 (IC50 = 61, 537, and 568 nM, respectively). VU0463271 exhibits much reduced efficacy against the related Na-K-2Cl cotransporter 1 (NKCC1 inhibition max ~37% at 100 μM) and little affinity toward a panel of 68 GPCRs, ion channels, and transporters. Although VU0463271 is reported to exhibit affinity toward mitochondrial translocator protein TSPO (IC50 of 204 nM against PK11195 binding; rat heart) and inhibitory potency against α1B adrenergic receptor (IC50 of 364.7 nM; human α1B CHO transfectants), these proteins are not known to affect chloride homeostasis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Subramanian et al.
Translational psychiatry, 8(1), 16-16 (2018-01-11)
Autism spectrum disorder (ASD) and temporal lobe epilepsy exhibit remarkable comorbidity, but for reasons not clearly understood. To reveal a common pathophysiological mechanism, we here describe and characterize an in vitro epileptiform activity in the rat hippocampus that exhibits common
Hannah Alfonsa et al.
Nature communications, 7, 13495-13495 (2016-11-18)
Chloride regulation affects brain function in many ways, for instance, by dictating the GABAergic reversal potential, and thereby influencing neuronal excitability and spike timing. Consistent with this, there is increasing evidence implicating chloride in a range of neurological conditions. Investigations
Matthew R Kelley et al.
Neuropharmacology, 108, 103-110 (2016-04-26)
Impaired neuronal inhibition has long been associated with the increased probability of seizure occurrence and heightened seizure severity. Fast synaptic inhibition in the brain is primarily mediated by the type A γ-aminobutyric acid receptors (GABAARs), ligand-gated ion channels that can
Eric Delpire et al.
Bioorganic & medicinal chemistry letters, 22(14), 4532-4535 (2012-06-26)
Further chemical optimization of the MLSCN/MLPCN probe ML077 (KCC2 IC(50)=537 nM) proved to be challenging as the effort was characterized by steep SAR. However, a multi-dimensional iterative parallel synthesis approach proved productive. Herein we report the discovery and SAR of
Peter M Klein et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 38(5), 1232-1248 (2017-12-24)
Maintenance of a low intracellular Cl- concentration ([Cl-]i) is critical for enabling inhibitory neuronal responses to GABAA receptor-mediated signaling. Cl- transporters, including KCC2, and extracellular impermeant anions ([A]o) of the extracellular matrix are both proposed to be important regulators of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service