Skip to Content
Merck
All Photos(1)

Key Documents

SML2074

Sigma-Aldrich

FX1

≥98% (HPLC)

Synonym(s):

(5Z)-5-(5-Chloro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)-4-oxo-2-thioxo-3-thiazolidinepropanoic acid, (Z)-3-(5-(5-Chloro-2-oxoindolin-3-ylidene)-4-oxo-2-thioxothiazolidin-3-yl)propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9ClN2O4S2
CAS Number:
Molecular Weight:
368.82
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

red-brown

storage temp.

2-8°C

SMILES string

S1\C(=C2\c3c(ccc(c3)Cl)NC\2=O)\C(=O)N(C1=S)CCC(=O)O

InChI key

VISHSGZPGQKEFX-KHPPLWFESA-N

Biochem/physiol Actions

FX1 is a BCL6 inhibitor that effectively blocks BCL6 N-terminal BTB domain-mediated corepressors chromosome recruitment (by 61-87%/SMRT and 67-82%/BCOR; 50 μM FX1 for 30 min) and selectively suppresses BCL6-depenent growth (GI50 16-54 μM; >125 μM against BCL6-independent cells) in diffuse large B cell lymphoma (DLBCL) cultures, exhibiting greater affinity than its structure analog 79-6 or BCL6 corepressor SMRT for BTB domain (KD = 7 μM/FX1, 30 μM/SMRT, 129 μM/79-6). FX1 shows greater efficacy than 79-6 in reversing BCL6/corepressors-mediated target genes repression in vitro (IC50 = 35 μM vs. 318 μM with 79-6 by HEK293T-based reporter assay) and in suppressing OCI-Ly7 DLBCL xenograft tumor growth mice in vivo (100% vs. 45% suppression with respective compound via 25 mg/kg/day i.p.).
Second generation 79-6 derived BCL6 inhibitor with improved efficacy against diffuse large B cell lymphoma (DLBCL) both in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wendy Béguelin et al.
Cancer cell, 30(2), 197-213 (2016-08-10)
The EZH2 histone methyltransferase mediates the humoral immune response and drives lymphomagenesis through formation of bivalent chromatin domains at critical germinal center (GC) B cell promoters. Herein we show that the actions of EZH2 in driving GC formation and lymphoma
William McCoull et al.
Journal of medicinal chemistry, 60(10), 4386-4402 (2017-05-10)
Inhibition of the protein-protein interaction between B-cell lymphoma 6 (BCL6) and corepressors has been implicated as a therapeutic target in diffuse large B-cell lymphoma (DLBCL) cancers and profiling of potent and selective BCL6 inhibitors are critical to test this hypothesis.
Mariano G Cardenas et al.
The Journal of clinical investigation, 126(9), 3351-3362 (2016-08-03)
Diffuse large B cell lymphomas (DLBCLs) arise from proliferating B cells transiting different stages of the germinal center reaction. In activated B cell DLBCLs (ABC-DLBCLs), a class of DLBCLs that respond poorly to current therapies, chromosomal translocations and amplification lead
H S Madapura et al.
Oncogene, 36(32), 4619-4628 (2017-04-04)
B-cell CLL/lymphoma 6 (BCL6) exerts oncogenic effects in several human hematopoietic malignancies including chronic myeloid leukemia (CML), where BCL6 expression was shown to be essential for CML stem cell survival and self-renewal during imatinib mesylate (IM) treatment. As several lines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service