Skip to Content
Merck
All Photos(1)

Key Documents

SML1853

Sigma-Aldrich

Kaempferitrin

≥97% (NMR)

Synonym(s):

3,7-Bis((6-deoxy-α-L-mannopyranosyl)oxy)-5-hydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Kaempferol 3,7-dirhamnoside, Lespedin, Lespenefril, Lespenephryl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O14
CAS Number:
Molecular Weight:
578.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

(Cinnamomum sp.)

Quality Level

Assay

≥97% (NMR)

form

powder

storage condition

desiccated
protect from light

color

white to beige

storage temp.

−20°C

SMILES string

OC(C=C1)=CC=C1C2=C(O[C@@H]3O[C@@H](C)[C@H](O)[C@@H](O)[C@H]3O)C(C4=C(O)C=C(O[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)C=C4O2)=O

InChI

1S/C27H30O14/c1-9-17(30)20(33)22(35)26(37-9)39-13-7-14(29)16-15(8-13)40-24(11-3-5-12(28)6-4-11)25(19(16)32)41-27-23(36)21(34)18(31)10(2)38-27/h3-10,17-18,20-23,26-31,33-36H,1-2H3/t9-,10-,17-,18-,20+,21+,22+,23+,26-,27-/m0/s1

InChI key

PUPKKEQDLNREIM-QNSQPKOQSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Kaempferitrin (Lespenefril) is a naturally-occurring kaempferol glycoside with antimicrobial, antioxidant, and anti-inflammatory activities.
Kaempferitrin is a flavonol glycoside, present in several plants, predominant in Bauhinia. It is known to stimulate insulin signaling pathway and induces glucose uptake, by mediating GLUT4 (glucose transporter 4) translocation to adipocyte membrane.. Kaempferitrin aids in lowering blood glucose level and therefore helps to improve the glycemic condition in diabetic animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaempferitrin activates the insulin signaling pathway and stimulates secretion of adiponectin in 3T3-L1 adipocytes
Tzeng YM, et al.
European Journal of Pharmacology, 607(1-3), 27-34 (2009)
Medicinal and Aromatic Plants of South America: Brazil, 142-142 (2018)
Xue-Qin Ma et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 22(13), 1159-1162 (2015-11-26)
Podocarpium podocarpum (DC.), an edible and medicinal plant popularly used for the treatment of bruises and fracture in Chinese folk medicine, has been proved to possess significant antiosteoporotic effect in our latest research. Our study aimed to investigate the in
Julia Cassani et al.
Molecules (Basel, Switzerland), 19(12), 21442-21461 (2014-12-24)
We evaluated the antidepressant-like effect of kaempferitrin (Km) isolated from the plant Justicia spicigera (Asteraceae), which is used in traditional medicine for relieving emotional disorders, such as "la tristeza" (sadness or dysthymia) and "el humor" (mood changes). The actions of
Discovery and Development of Antidiabetic Agents from Natural Products, 118-118 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service