Skip to Content
Merck
All Photos(1)

Documents

SML1537

Sigma-Aldrich

Cantharidin Ready Made Solution

0.1 mg/mL in DMSO

Synonym(s):

3a,7a-Dimethylhexahydro-4,7-epoxyisobenzofuran-1,3-dione, 7a-Dimethylhexahydro-3a,4,7-epoxyisobenzofuran, Cantharidine, Hexahydro-3a,7a-dimethyl-4,7-epoxyisobenzofuran-1,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O4
CAS Number:
Molecular Weight:
196.20
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

insect

Quality Level

form

solution

concentration

0.1 mg/mL in DMSO

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C10H12O4/c1-9-5-3-4-6(13-5)10(9,2)8(12)14-7(9)11/h5-6H,3-4H2,1-2H3/t5-,6+,9+,10-

InChI key

DHZBEENLJMYSHQ-XCVPVQRUSA-N

Biochem/physiol Actions

It is a component of mylabris, a traditional Chinese medicine. Cantharidin exhibits anti-tumor activity. In addition, cantharidin can work against multidrug-resistant cells. In the epidermis, it causes release of neutral serine proteases, leading to degeneration of the desmosmal plaque. This results in acantholysis and intraepidermal blistering as well as skin lysis. PP2A is involved in the cell cycle, cell death, growth and cell-fate determination.
Cantharidin inhibits protein phosphatase 2A.

Other Notes

Stable for at least two years at 2-8° C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Li et al.
Cancer science, 101(5), 1226-1233 (2010-03-25)
Cantharidin is an active constituent of mylabris, a traditional Chinese medicine. It is a potent and selective inhibitor of protein phosphatase 2A (PP2A) that plays an important role in control of cell cycle, apoptosis, and cell-fate determination. Owing to its
Rolf Rauh et al.
Chinese medicine, 2, 8-8 (2007-07-06)
Herbal medicine is one of the forms of traditional medical practice. Traditional Chinese medicine (TCM) and traditional Vietnamese medicine (TVM) are well-known for their long-standing tradition of herbal medicine. Secreted by many species of blister beetle, most notably by the
L Moed et al.
Archives of dermatology, 137(10), 1357-1360 (2001-10-27)
Cantharidin, a vesicant produced by beetles in the order Coleoptera, has a long history in both folk and traditional medicine. In dermatology, topical cantharidin has long been used to treat warts and molluscum. In 1962, cantharidin lost Food and Drug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service