Skip to Content
Merck
All Photos(1)

Key Documents

SML1490

Sigma-Aldrich

Pemetrexed disodium heptahydrate

≥98% (HPLC)

Synonym(s):

HSDB 7316, LY 231,514, LY 231514, N-(4-(2-(2-Amino-3,4-dihydro-4-oxo-7H-pyrrolo(2,3-d)pyrimdin-5-yl)ethyl)benzoyl)glutamic acid sodium salt hydrate, N-(4-(2-(2-Amino-4,7-dihydro-4-oxo-1H-pyrrolo(2,3-d)pyrimidin-5-yl)ethyl)benzoyl)-L-glutamic acid sodium salt hydrate, UNII-04Q9AIZ7NO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19N5O6 · 2Na · 7H2O
CAS Number:
Molecular Weight:
597.48
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 30 mg/mL, clear

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[nH]1c2[nH]cc(c2[c](nc1N)=O)CCc3ccc(cc3)C(=O)N[C@@H](CCC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.O

InChI

1S/C20H21N5O6.2Na.7H2O/c21-20-24-16-15(18(29)25-20)12(9-22-16)6-3-10-1-4-11(5-2-10)17(28)23-13(19(30)31)7-8-14(26)27;;;;;;;;;/h1-2,4-5,9,13H,3,6-8H2,(H,23,28)(H,26,27)(H,30,31)(H4,21,22,24,25,29);;;7*1H2/q;2*+1;;;;;;;/p-2/t13-;;;;;;;;;/m0........./s1

InChI key

QJVSMHJWAOSBMD-MYXYZBIASA-L

Application

Pemetrexed disodium heptahydrate has been used as a folate antimetabolite to study its effects on ovarian clear cell carcinoma cells. It has also been used as an antifolate to determine its effects on neuroblastoma cell lines.

Biochem/physiol Actions

Pemetrexed exhibits its activity against several tumors such as colorectal, breast, gastric, pancreatic, and cervical cancer. It acts as a substrate for multidrug resistance protein transporters.
Pemetrexed is antifolate cancer drug approved for treatment of variety of cancer including pleural mesothelioma and non-small cell lung cancer.
Pemetrexed is antifolate cancer drug approved for treatment of variety of cancer including pleural mesothelioma and non-small cell lung cancer. Pemetrexed is a folate antimetabolite that inhibits thymidylate synthase, dihydrofolate reductase and glycinamide ribonucleotide formyltransferase, three enzymes used in purine and pyrimidine synthesis.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 1A - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood,Immune system,Testes

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alex A Adjei
Clinical cancer research : an official journal of the American Association for Cancer Research, 10(12 Pt 2), 4276s-4280s (2004-06-26)
Pemetrexed (ALIMTA, LY231514, MTA) is a novel antimetabolite that inhibits at least three enzymes involved in the folate pathway. These enzymes are thymidylate synthase, dihydrofolate reductase, and glycinamide ribonucleotide formyltransferase. Pemetrexed has demonstrated clinical activity in non-small cell lung cancer
Ning Mu et al.
Cell death & disease, 14(9), 645-645 (2023-10-01)
Brain expressed X-linked gene 2 (BEX2) encoded protein was originally identified to promote transcription by interacting with several transcription factors in the DNA-binding complexes. Recently, BEX2 was found to be localized in cytosol and/or mitochondria and regulate apoptosis in cancer
Pooyeh Farahmand et al.
Frontiers in toxicology, 5, 1200650-1200650 (2023-07-13)
Hypothesis: Asbestos-driven inflammation contributes to malignant pleural mesothelioma beyond the acquisition of rate-limiting mutations. Methods: Genetically modified conditional allelic mice that were previously shown to develop mesothelioma in the absence of exposure to asbestos were induced with lentiviral vector expressing
Ping Tian et al.
PloS one, 19(2), e0298295-e0298295 (2024-02-14)
Timeless is well-known for its key role in replication checkpoints. Recent studies reveal the involvement of Timeless and specificity protein (SP) 1 in human malignancies. However, no evidence proved the interaction between SP3 and Timeless in lung adenocarcinoma (LUAD). The
Ken Yamashita et al.
Cancer science, 111(7), 2431-2439 (2020-05-18)
MYCN gene amplification is consistently associated with poor prognosis in patients with neuroblastoma, a pediatric tumor arising from the sympathetic nervous system. Conventional anticancer drugs, such as alkylating agents and platinum compounds, have been used for the treatment of high-risk

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service