Skip to Content
Merck
All Photos(1)

Documents

SML1122

Sigma-Aldrich

BTB06584

≥98% (HPLC)

Synonym(s):

2-Nitro-5-(phenylsulfonyl)phenyl 4-chloro-benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H12ClNO6S
CAS Number:
Molecular Weight:
417.82
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(C1=CC=C([N+]([O-])=O)C(OC(C2=CC=C(Cl)C=C2)=O)=C1)(C3=CC=CC=C3)=O

InChI

1S/C19H12ClNO6S/c20-14-8-6-13(7-9-14)19(22)27-18-12-16(10-11-17(18)21(23)24)28(25,26)15-4-2-1-3-5-15/h1-12H

InChI key

WNDWKKPBLAKXMI-UHFFFAOYSA-N

Biochem/physiol Actions

BTB06584 inhibits the ATP hydrolase activity of mitochondrial F1F0 ATP synthase. The compound BTB06584 has no affect on oxygen consumption or mitochondrial membrane potential in HL-1, a mouse cardiac cell line, but blocks ATP consumption and ischemic cell death following inhibition of cellular respiration.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service