Skip to Content
Merck
All Photos(1)

Key Documents

SML1121

Sigma-Aldrich

SB743921 hydrochloride

≥98% (HPLC)

Synonym(s):

N-(3-Aminopropyl)-N-[(1R)-1-[7-chloro-4-oxo-3-(phenylmethyl)-4H-1-benzopyran-2-yl]-2-methylpropyl]-4-methylbenzamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H33ClN2O3 · HCl
CAS Number:
Molecular Weight:
553.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 5 mg/mL, clear

storage temp.

−20°C

SMILES string

CC(C)[C@@H](N(CCCN)C(C1=CC=C(C)C=C1)=O)C2=C(CC3=CC=CC=C3)C(C4=C(O2)C=C(Cl)C=C4)=O.Cl

InChI

1S/C31H33ClN2O3.ClH/c1-20(2)28(34(17-7-16-33)31(36)23-12-10-21(3)11-13-23)30-26(18-22-8-5-4-6-9-22)29(35)25-15-14-24(32)19-27(25)37-30;/h4-6,8-15,19-20,28H,7,16-18,33H2,1-3H3;1H/t28-;/m1./s1

InChI key

MLMZVWABFOLFGV-LNLSOMNWSA-N

Biochem/physiol Actions

SB743921 is an inhibitor of kinesin spindle protein (KSP) with Ki of 0.1 nM, almost no affinity to MKLP1, Kin2, Kif1A, Kif15, KHC, Kif4, and CENP-E.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehdi Bouhaddou et al.
Cell, 182(3), 685-712 (2020-07-10)
The causative agent of the coronavirus disease 2019 (COVID-19) pandemic, severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), has infected millions and killed hundreds of thousands of people worldwide, highlighting an urgent need to develop antiviral therapies. Here we present a

Articles

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service