Skip to Content
Merck
All Photos(2)

Key Documents

SML0700

Sigma-Aldrich

FTY720

≥98% (HPLC), powder, sphingosine 1-phosphate receptor modulator

Synonym(s):

2-Amino-2-[2-(4-octyl-phenyl)-ethyl]-propane-1,3-diol hydrochloride, Fingolimod hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H33NO2 · HCl
CAS Number:
Molecular Weight:
343.93
UNSPSC Code:
12352211
NACRES:
NA.77

Product Name

FTY720, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

water: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

Cl.NC(CO)(CO)CCc1ccc(cc1)CCCCCCCC

InChI

1S/C19H33NO2.ClH/c1-2-3-4-5-6-7-8-17-9-11-18(12-10-17)13-14-19(20,15-21)16-22;/h9-12,21-22H,2-8,13-16,20H2,1H3;1H

InChI key

SWZTYAVBMYWFGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

FTY720 has been used as an immunomodulator and is used to investigate its effect on in vitro gastrulation model of P19C5 stem cells. It is also used to determine its efficacy in prolonging the survival corneal transplantation. FTY720 has been used to study its effect on the proliferation and differentiation of cultured embryonic hippocampal neural stem cells (NSCs) using the 5-bromo-2-deoxyuridine incorporation assay, the neurosphere formation assay and western blot analysis.

Biochem/physiol Actions

FTY720 is an immunomodulating drug and sphingosine 1-phosphate (S1P) receptor modulator. Phosphorylation of FTY270 by sphingosine kinase causes S1P1R internalization, which sequesters lymphocytes in lymph nodes, preventing them from taking part in an autoimmune response. Clinically, it has been approved for the treatment of multiple sclerosis (MS). It has also been shown to block and reverse paclitaxel-induced chemotherapy induced peripheral neuropathy (CIPN) through S1PR inhibition as well as inhibit the activity of histone deacetylases in the hippocampus of mouse brains, thereby modulating memory.
FTY720, an immunomodulating drug, acts as a sphingosine 1-phosphate (S1P) receptor modulator.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2

Target Organs

Immune system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Goeril Karlsson et al.
Neurology, 82(8), 674-680 (2014-01-28)
To report outcomes of pregnancies that occurred during the fingolimod clinical development program. Pregnancy outcomes from phase II, phase III, and phase IV clinical studies (with optional extensions) were reported by clinical trial investigators. Fingolimod exposure in utero was defined
M Mehling et al.
Neurology, 76(8 Suppl 3), S20-S27 (2011-02-26)
The oral sphingosine 1-phosphate (S1P) receptor (S1PR) modulator fingolimod has been shown to be effective in the treatment of patients with relapsing multiple sclerosis (MS). The drug binds with high affinity to 4 of the 5 G-protein-coupled S1P receptors (S1P(1-5)).
Paroxysmal atrial fibrillation after initiation of fingolimod for multiple sclerosis treatment.
Linda Rolf et al.
Neurology, 82(11), 1008-1009 (2014-02-14)
Fingolimod and teriflunomide attenuate neurodegeneration in mouse models of neuronal ceroid lipofuscinosis.
Groh J, et al.
Molecular Therapy, 25(8), 1889-1899 (2017)
The sphingosine-1-phosphate analogue, FTY-720, promotes the proliferation of embryonic neural stem cells, enhances hippocampal neurogenesis and learning and memory abilities in adult mice.
Sun Y, et al.
British Journal of Pharmacology, 173(18), 2793-2807 (2016)

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service