Skip to Content
Merck
All Photos(1)

Documents

SML0547

Sigma-Aldrich

Loteprednol Etabonate

≥98% (HPLC)

Synonym(s):

(11b,17a)-17-[(Ethoxycarbonyl)oxy]-11-hydroxy-3-oxo-Androsta-1,4-diene-17-carboxylic acid chloromethyl ester, CDDD 5604, HGP 1, P 5604

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31ClO7
CAS Number:
Molecular Weight:
466.95
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

CCOC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)OCCl

InChI

1S/C24H31ClO7/c1-4-30-21(29)32-24(20(28)31-13-25)10-8-17-16-6-5-14-11-15(26)7-9-22(14,2)19(16)18(27)12-23(17,24)3/h7,9,11,16-19,27H,4-6,8,10,12-13H2,1-3H3/t16-,17-,18-,19+,22-,23-,24-/m0/s1

InChI key

DMKSVUSAATWOCU-HROMYWEYSA-N

Gene Information

human ... NR3C1(2908)

Biochem/physiol Actions

Loteprednol Etabonate is a "soft" steroid, typically used in topical applications for inflammatory conditions of the eye. Loteprednol Etabonate is an agonist of both glucocorticoid and mineralcorticoid receptors.
Loteprednol Etabonate is an anti-inflammatory corticosteroid (ophthalmology).
Loteprednol etabonate (LE) is a key site-active corticosteroid. It is produced by structural modifications of prednisolone-related compounds to form an inactive metabolite. Double-masked study states that loteprednol etabonate is used to treat giant papillary conjunctivitis, seasonal allergic conjunctivitis, postoperative inflammation and uveitis. LE has good ocular permeation properties.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edoardo Villani et al.
Optometry and vision science : official publication of the American Academy of Optometry, 92(9), e290-e295 (2015-04-25)
To evaluate, by in vivo laser scanning confocal microscopy (LSCM), the corneal findings in moderate-to-severe dry eye patients before and after treatment with topical corticosteroid and to associate the confocal findings to the clinical response. Fifty eyes of 50 patients
Maamoun Abdul Fattah et al.
Journal of refractive surgery (Thorofare, N.J. : 1995), 36(8), 498-505 (2020-08-14)
To evaluate the efficacy of simultaneous laser in situ keratomileusis (LASIK) and small-aperture corneal inlay (KAMRA; AcuFocus, Inc) implantation in hyperopic presbyopic eyes at 5 years postoperatively. This was a retrospective single-center study of patients with hyperopia and presbyopia who
Ocular absorption and distribution of loteprednol etabonate, a soft steroid, in rabbit eyes
Druzgala P, et al.
Current Eye Research, 10(10), 933-937 (1991)
Kaiwei Cao et al.
Eye and vision (London, England), 7, 36-36 (2020-07-14)
Several planning algorithms have been developed for topography-guided custom ablation treatment (T-CAT), but each has its own deficiencies. The purpose of this study is to demonstrate the potential of a novel mutual comparative analysis (MCA) informed by manifest refraction and
Midhat H Abdulreda et al.
Diabetologia, 62(7), 1237-1250 (2019-05-16)
Autoimmune attack against the insulin-producing beta cells in the pancreatic islets results in type 1 diabetes. However, despite considerable research, details of the type 1 diabetes immunopathology in situ are not fully understood mainly because of difficult access to the

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service