Skip to Content
Merck
All Photos(1)

Documents

SML0512

Sigma-Aldrich

BLT-4

≥98% (HPLC)

Synonym(s):

INF 271, MIT 9952-29, N-(2-Methoxyphenyl)-N′-2-naphthalenyl-urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N2O2
CAS Number:
Molecular Weight:
292.33
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL (clear solution)

storage temp.

room temp

SMILES string

COc1ccccc1NC(=O)Nc2ccc3ccccc3c2

InChI

1S/C18H16N2O2/c1-22-17-9-5-4-8-16(17)20-18(21)19-15-11-10-13-6-2-3-7-14(13)12-15/h2-12H,1H3,(H2,19,20,21)

InChI key

ISADADAHPHHLEM-UHFFFAOYSA-N

Biochem/physiol Actions

BLT-4 is a blocker of lipid transport-4 (BLT-4). BLT-4 is a specific, reversible inhibitor of scavenger receptor, class B, type I (SR-BI) mediated lipid transfer. The molecule blocks the cellular uptake and transfer of cholesterol ester from HDL.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elsa Westerlund et al.
Frontiers in immunology, 10, 1385-1385 (2019-07-06)
The common human pathogen Group A Streptococcus (GAS) causes superficial as well as invasive, life-threatening diseases. An increase in the occurrence of invasive GAS infection by strains of the M1 and M89 serotypes has been correlated with increased expression of
Guillemette Maurin et al.
AIDS (London, England), 29(9), 1025-1033 (2015-07-01)
Chronic hepatitis C virus (HCV) infection causes severe liver disease in HIV-infected patients and liver transplant recipients. The impact of serum and immunoglobulin on viral entry was analysed in these patients. Sera from 60 anti-HCV positive patients, including 30 who
Thomas J F Nieland et al.
Journal of lipid research, 45(7), 1256-1265 (2004-04-23)
Scavenger receptor class B type I (SR-BI) and ABCA1 are structurally dissimilar cell surface proteins that play key roles in HDL metabolism. SR-BI is a receptor that binds HDL with high affinity and mediates both the selective lipid uptake of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service