Skip to Content
Merck
All Photos(1)

Key Documents

SML0494

Sigma-Aldrich

VU0361737

≥98% (HPLC)

Synonym(s):

CID-44191096, ML128, N-(4-Chloro-3-methoxyphenyl)-2-pyridinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11ClN2O2
CAS Number:
Molecular Weight:
262.69
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

film

color

white

solubility

DMSO: 15 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

COc1cc(NC(=O)c2ccccn2)ccc1Cl

InChI

1S/C13H11ClN2O2/c1-18-12-8-9(5-6-10(12)14)16-13(17)11-4-2-3-7-15-11/h2-8H,1H3,(H,16,17)

InChI key

ARYUXFNGXHNNDM-UHFFFAOYSA-N

Biochem/physiol Actions

VU0361737 is an mGluR4-specific positive allosteric modulator (PAM) with greater than 50 fold selectivity over other mGluR subtypes. The EC50 values for human and rat mGlu4 are 240 and 110 nM, respectively, compared to greater than 10 mM for all other mGluRs.
VU0361737 is an mGluR4-specific positive allosteric modulator (PAM).

Features and Benefits

This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.

Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.

Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.

Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service