Skip to Content
Merck
All Photos(1)

Key Documents

SML0338

Sigma-Aldrich

Lornoxicam

≥98% (HPLC)

Synonym(s):

6-Chloro-4-hydroxy-2-methyl-N-2-pyridinyl-2H-thieno[2,3-e]-1,2-thiazine-3-carboxamide 1,1-dioxide, Chlortenoxicam, Ro 13-9297, TS 110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10ClN3O4S2
CAS Number:
Molecular Weight:
371.82
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

faintly yellow to dark yellow

solubility

DMSO: >5 mg/mL (warmed)

storage temp.

−20°C

SMILES string

CN1C(C(=O)Nc2ccccn2)=C(O)c3sc(Cl)cc3S1(=O)=O

InChI

1S/C13H10ClN3O4S2/c1-17-10(13(19)16-9-4-2-3-5-15-9)11(18)12-7(23(17,20)21)6-8(14)22-12/h2-6,18H,1H3,(H,15,16,19)

InChI key

WLHQHAUOOXYABV-UHFFFAOYSA-N

General description

Lornoxicam belongs to the oxicam class. It has anti-inflammatory and antipyretic properties. Lornoxicam prevents the synthesis of prostaglandin (PG) by inhibiting cyclo-oxygenase. It is used to relieve various types of symptoms associated with osteoarthritis, rheumatoid arthritis, ankylosing spondylitis, acute sciatica and low back pain.

Application

Lornoxicam has been used as a drug in melanin binding study with cassette dosing and rapid equilibrium dialysis inserts.

Biochem/physiol Actions

Lornoxicam is an oxicam-class NSAID with strong analgesic properties.
Lornoxicam is an oxicam-class NSAID; cyclooxygenase inhibitor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianjun Zhang et al.
Pakistan journal of pharmaceutical sciences, 25(2), 371-375 (2012-03-31)
A rapid, isocratic stability indicating high performance liquid chromatographic method was developed and validated for the estimation of lornoxicam in its powder for injection. The analysis was performed on a Shimadzu VP-ODS (4. 6 mm x 15 cm, 5 µm)
M Arslan et al.
Bratislavske lekarske listy, 113(4), 211-213 (2012-04-17)
Lornoxicam and iv paracetamol are commonly preferred to be used for postoperative analgesia. Although Aspirin is a well known non-steroid anti-inflammatory drug that decreases the erythrocyte deformability, there is no study comparing lornoxicam and iv paracetamol regarding their effects on
Mustafa Arslan et al.
Agri : Agri (Algoloji) Dernegi'nin Yayin organidir = The journal of the Turkish Society of Algology, 23(4), 160-166 (2012-02-01)
The purpose of the present study was to determine the efficacy of intravenous (iv) paracetamol and iv lornoxicam on postoperative analgesia and the reduction in tramadol consumption. Sixty patients (ASA class 1-2, age: 18-72 years) undergoing thyroidectomy were enrolled in
Susanne Schroeder et al.
Rheumatology international, 32(9), 2661-2667 (2011-07-27)
The local tolerability of lornoxicam (Xefo) after single and repeated intraarticular administration was assessed in the rabbit and compared to established standard therapies (hyaluronic acid--Synvisc and the glucocorticoid triamcinolone--Triam), and the results are discussed in the context of the literature.
Hany A Mowafi et al.
World journal of surgery, 36(9), 2039-2044 (2012-05-16)
The aim of this prospective, randomized, double-blind study was to determine the more effective supplemental analgesic, paracetamol or lornoxicam, for postoperative pain relief after lower abdominal surgery. Sixty patients scheduled for lower abdominal surgery under general anesthesia were randomly allocated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service