Skip to Content
Merck
All Photos(1)

Key Documents

SML0293

Sigma-Aldrich

Telbivudine

≥98% (HPLC)

Synonym(s):

β-L-Thymidine, 1-(2-Deoxy-β-L-erythro-pentofuranosyl)-5-methyl-2,4(1H,3H)-Pyrimidinedione, 2′-Deoxy-L-thymidine, Epavudine, L-Thymidine, NV 02B, Sebivo

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O5
CAS Number:
Molecular Weight:
242.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -15 to -22°, c = 1 in H2O

color

white to beige

solubility

H2O: 10 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

CC1=CN([C@@H]2C[C@@H](O)[C@H](CO)O2)C(=O)NC1=O

InChI

1S/C10H14N2O5/c1-5-3-12(10(16)11-9(5)15)8-2-6(14)7(4-13)17-8/h3,6-8,13-14H,2,4H2,1H3,(H,11,15,16)/t6-,7+,8+/m1/s1

InChI key

IQFYYKKMVGJFEH-CSMHCCOUSA-N

Application

Telbivudine has been used to explore the mechanisms of telbivudine (LdT) induced creatine kinase (CK) elevation.

Biochem/physiol Actions

Telbivudine is a nucleoside analog that is widely used to treat hepatitis B virus (HBV) infection.
Telbivudine is a synthetic molecule containing β-L-configuration. Telbivudine improves the estimated glomerular filtration rate (eGFR) in response to chronic hepatitis B (CHB). It prevents perinatal transmission and enhances the hepatitis B e antigen (HBeAg) seroconversion. It is also known to cause side effects such as creatine kinase (CK) elevation and fatal rhabdomyolysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weiting Qin et al.
PloS one, 8(10), e75840-e75840 (2013-10-12)
Sepsis continues to be a challenge in clinic. The rates of mortality in sepsis patients remain high. The present study aimed to investigate the effects and the underlying mechanisms of carbon monoxide-releasing molecules II (CORM-2)-liberated CO on suppressing inflammatory response
Sophie Van Linthout et al.
ESC heart failure, 5(5), 818-829 (2018-08-14)
Myocarditis is often associated with parvovirus B19 (B19V) persistence, which can induce vascular damage. Based on the antiviral and anti-inflammatory properties of telbivudine, we aimed to evaluate its efficacy to protect B19V-infected endothelial cells in vitro and to treat chronic
Ailin Chen et al.
Cancer communications (London, England), 41(2), 121-139 (2021-01-21)
Overexpression of Aurora-A (AURKA) is a feature of breast cancer and associates with adverse prognosis. The selective Aurora-A inhibitor alisertib (MLN8237) has recently demonstrated promising antitumor responses as a single agent in various cancer types but its phase III clinical
Quanxin Wu et al.
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association, 13(6), 1170-1176 (2014-09-25)
Hepatitis B virus (HBV) infection is a leading cause of liver diseases. We investigated the efficacy and safety of telbivudine in preventing transmission of HBV from hepatitis B e antigen-positive pregnant women with high viral loads to their infants in
Faezeh Koohestani et al.
PloS one, 8(9), e75844-e75844 (2013-09-17)
Uterine leiomyomas (ULs) are benign tumors occurring in the majority of reproductive aged women. Despite the high prevalence of these tumors, little is known about their etiology. A hallmark of ULs is the excessive deposition of extracellular matrix (ECM), primarily

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service