Skip to Content
Merck
All Photos(1)

Key Documents

SML0278

Sigma-Aldrich

SR 48692

≥98% (HPLC)

Synonym(s):

2-[[[1-(7-Chloro-4-quinolinyl)-5-(2,6-dimethoxyphenyl)-1H-pyrazol-3-yl]carbonyl]amino]-tricyclo[3.3.1.13,7]decane-2-carboxylic acid, Meclinertant

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H31ClN4O5
CAS Number:
Molecular Weight:
587.07
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥2 mg/mL (warmed)

storage temp.

−20°C

SMILES string

COc1cccc(OC)c1-c2cc(nn2-c3ccnc4cc(Cl)ccc34)C(=O)NC5(C6CC7CC(C6)CC5C7)C(O)=O

InChI

1S/C32H31ClN4O5/c1-41-27-4-3-5-28(42-2)29(27)26-16-24(36-37(26)25-8-9-34-23-15-21(33)6-7-22(23)25)30(38)35-32(31(39)40)19-11-17-10-18(13-19)14-20(32)12-17/h3-9,15-20H,10-14H2,1-2H3,(H,35,38)(H,39,40)

InChI key

DYLJVOXRWLXDIG-UHFFFAOYSA-N

Gene Information

human ... NTSR1(4923)

Application

SR 48692 has been used as a neurotensin high-affinity receptor 1 (NTSR1) antagonist:
  • to explore the function of NTSR1 in glioblastoma (GBM) cells
  • to determine the roles of neurotensin (NT) in the regulation of bile acid (BA) uptake, in vivo
  • to explore the involvement of NTSR1 versus NTSR2 in mice

Biochem/physiol Actions

SR 48692 is a high affinity, orally bioavailable and selective nonpeptide NT1 neurotensin receptor antagonist that antagonizes neurotensin-induced calcium mobilization with a pA2 of 8.13 in HT-29 human colon carcinoma cell line, and blocks the ability of neurotensin to increase GABA levels in the prefrontal cortex.

Features and Benefits

This compound is featured on the Neurotensin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristóf László et al.
Behavioural brain research, 226(2), 597-600 (2011-09-29)
Tridecapeptide neurotensin (NT) acts as a neurotransmitter and/or neuromodulator and plays a role in learning and reinforcement. The central nucleus of amygdala (CeA), which is relatively rich in NT and neurotensin-1 receptors (NTS1), participates in the regulation of memory and
Fabiana G Costa et al.
Neuropeptides, 41(2), 83-91 (2007-02-06)
Psychostimulant-induced locomotor sensitization has been related to changes within the mesolimbic dopamine system and has been suggested to be useful to study mechanisms underlying drug craving. Neurotensin is a neuropeptide co-localized with dopamine in the mesolimbic system. The response to
James B Thomas et al.
Bioorganic & medicinal chemistry letters, 19(5), 1438-1441 (2009-02-07)
In a search for nonpeptide agonists for the neurotensin receptor (NTR1), we replaced the adamantyl amino acid moiety found in the antagonist SR48692 (1a) with leucine and related alpha-alkylamino acids found in peptide agonists. When tested in a calcium mobilization
C Jomphe et al.
The European journal of neuroscience, 24(10), 2789-2800 (2006-11-23)
Several lines of evidence suggest a close association between dopamine (DA) and neurotensin (NT) systems in the CNS. Indeed, in the rodent brain, abundant NT-containing fibres are found in DA-rich areas such as the ventral tegmental area and substantia nigra.
Kristóf László et al.
Behavioural brain research, 208(2), 430-435 (2009-12-29)
In the central nervous system neurotensin (NT) acts as a neurotransmitter and neuromodulator. It was shown that NT has positive reinforcing effects after its direct microinjection into the ventral tegmental area. The central nucleus of amygdala (CeA), part of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service