Skip to Content
Merck
All Photos(1)

Key Documents

SML0099

Sigma-Aldrich

Fluocinonide

≥98% (HPLC)

Synonym(s):

(6α,11β,16α)-21-(acetyloxy)-6,9-difluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-pregna-1,4-diene-3,20-dione, Lonide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32F2O7
CAS Number:
Molecular Weight:
494.52
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +75 to +90°, c = 1 in dichloromethane

color

white to tan

solubility

DMSO: ≥19 mg/mL

storage temp.

2-8°C

SMILES string

CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]23C

InChI

1S/C26H32F2O7/c1-13(29)33-12-20(32)26-21(34-22(2,3)35-26)10-15-16-9-18(27)17-8-14(30)6-7-23(17,4)25(16,28)19(31)11-24(15,26)5/h6-8,15-16,18-19,21,31H,9-12H2,1-5H3/t15-,16-,18-,19-,21+,23-,24-,25-,26+/m0/s1

InChI key

WJOHZNCJWYWUJD-IUGZLZTKSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Flucinonide is a fluorinated glucocorticid that is used as a topical anti-inflammatory agent. Recently, flucinonide and other fluorinated glucocorticolids were identified as smoothend agonists. Flucinonide induces expression of Gli-reporter luciferase in Shh-LIGHT2 cells.
Fluocinonide has been used to study its effect on T-cell proliferation.
Fluocinonide is shown to be effective in the treatment of atopic dermatitis and oral lichen planus (OLP).

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Małgorzata Radwan-Oczko
Advances in clinical and experimental medicine : official organ Wroclaw Medical University, 22(6), 893-898 (2014-01-17)
Oral lichen planus is a common, chronic mucosal disease associated with a cell-mediated immunological dysfunction. The clinical manifestation is different when various forms, white and red, are considered. Erosive, atrophic, ulcerative lesions require long-term treatment, because of inflammation and severe
Emily Hix et al.
Dermatology online journal, 19(10), 20029-20029 (2013-10-22)
Adherence in the treatment of chronic inflammatory skin diseases such as atopic dermatitis is poor. Methods to improve adherence have proven difficult. To determine whether a short course of treatment with a high-potency corticosteroid will improve adherence compared to longer
Localized acral pityriasis lichenoides chronica: report of a case.
Jennifer J Halbesleben et al.
The Journal of dermatology, 38(8), 832-834 (2011-03-01)
Matthew T Woods et al.
Journal of drugs in dermatology : JDD, 10(2), 171-176 (2011-02-02)
To determine the effect a novel formulation of fluocinonide cream on skin barrier function in subjects with atopic dermatitis. The authors performed an open-label, investigator-blinded, side-by-side, controlled trial examining skin barrier function before and after a two-week course of a
Tinea incognito.
Rhonda Lesniak
Dermatology nursing, 20(5), 403-404 (2008-12-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service