Skip to Content
Merck
All Photos(1)

Key Documents

SMB00506

Sigma-Aldrich

1-Heptadecanoyl-rac-glycerol

≥98% (HPLC)

Synonym(s):

1-Margaroyl-rac-glycerol, Monoheptadecanoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H40O4
CAS Number:
Molecular Weight:
344.53
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

functional group

ketone

lipid type

neutral glycerides

shipped in

ambient

storage temp.

2-8°C

SMILES string

OCC(O)COC(CCCCCCCCCCCCCCCC)=O

InChI

1S/C20H40O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-20(23)24-18-19(22)17-21/h19,21-22H,2-18H2,1H3

InChI key

SVUQHVRAGMNPLW-UHFFFAOYSA-N

General description

1-Heptadecanoyl-rac-glycerol (2,3-dihydroxypropyl heptadecanoate) is a monoglyceride. It has been identified as one of the lipophilic compounds present in the woods of the different eucalypt hybrids and in the extracts of wheat bran.

Application

1-Heptadecanoyl-rac-glycerol may be employed as an internal standard during the quantitative estimation of 2-arachidonoyl glycerol (2-AG) in tissue lipid extracts of overnight fasted mice by LC/MS (Liquid chromatography–mass spectrometry). It may be employed as a neutral lipid standard for the analysis of cellular neutral lipid (such as cholesterol, cholesterol esters, wax esters) by HPLC.
Standard for the quantification and characterization of lipid mixtures

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David A Sullivan et al.
Investigative ophthalmology & visual science, 55(6), 3866-3877 (2014-05-29)
We hypothesize that culturing immortalized human meibomian gland epithelial cells in serum-containing medium will induce their differentiation. The purpose of this investigation was to begin to test our hypothesis, and explore the impact of serum on gene expression and lipid
Morphological characteristics and composition of lipophilic extractives and lignin in Brazilian woods from different eucalypt hybrids.
Prinsen P, et al.
Industrial Crops and Products, 36(1), 572-583 (2012)
Pepijn Prinsen et al.
Journal of agricultural and food chemistry, 62(7), 1664-1673 (2014-01-24)
Wheat bran, the major side-stream generated in the milling of wheat grains in the production of white flour, contains significant quantities of carbohydrate and proteins. While not interfering with flour utilization, the bran could be considered as an important feedstock
Takahiro Ogawa et al.
Rapid communications in mass spectrometry : RCM, 31(11), 928-936 (2017-04-04)
The acylglycerols in miso have not been studied although it is known that they are important to the taste. In order to determine the fatty acid constituents in the acylglycerols and analyze them individually, multiple reaction monitoring (MRM) was performed
U Taschler et al.
British journal of pharmacology, 172(17), 4419-4429 (2015-06-16)
Monoglyceride lipase (MGL) degrades 2-arachidonoyl glycerol (2-AG), an endogenous agonist of cannabinoid receptors (CB1/2 ). Because the CB1 receptor is involved in the control of gut function, we investigated the effects of pharmacological inhibition and genetic deletion of MGL on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service