Skip to Content
Merck
All Photos(2)

Key Documents

SMB00388

Sigma-Aldrich

2,4,3′,5′-tetramethoxystilbene

≥98% (HPLC)

Synonym(s):

2,4,3′,5′-tetramethoxystilbene, 1-[(1E)-2-(3,5-dimethoxyphenyl)ethenyl]-2,4-dimethoxy-benzene, 2,3′,4,5′-Tetramethoxystilbene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20O4
CAS Number:
Molecular Weight:
300.35
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.79

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white to yellow

storage temp.

−20°C

SMILES string

COC1=CC(OC)=CC=C1/C=C/C2=CC(OC)=CC(OC)=C2

InChI

1S/C18H20O4/c1-19-15-8-7-14(18(12-15)22-4)6-5-13-9-16(20-2)11-17(10-13)21-3/h5-12H,1-4H3/b6-5+

InChI key

JDBCWSHYEQUBLW-AATRIKPKSA-N

Biochem/physiol Actions

2,3′,4,5′-Tetramethoxystilbene, also known as TMS, is a selective and competitive inhibitor of cytochrome P450 1B. It is a synthetic trans-stilbene analog. TMS is an analog of resveratrol and is an anti-cancer agent. TMS is cytotoxic to gefitinib-resistant (G-R) non-small-cell lung cancer (NSCLC) cells. It binds to sarco/endoplasmic reticulum Ca2+-ATPase (SERCA) and promotes apoptosis and autophagy. TMS is also a selective inhibitor of CYP1B1.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modulation of human cytochrome P450 1B1 expression by 2, 4, 3?, 5?-tetramethoxystilbene
Chun YJ, et al.
Drug Metabolism and Disposition, 33(12), 1771-1776 (2005)
(Z) 3, 4, 5, 4?-trans-tetramethoxystilbene, a new analogue of resveratrol, inhibits gefitinb-resistant non-small cell lung cancer via selectively elevating intracellular calcium level
Fan XX, et al.
Scientific Reports, 5, 16348-16348 (2015)
The resveratrol analogue, 2, 3?, 4, 5?-tetramethoxystilbene, does not inhibit CYP gene expression, enzyme activity and benzo [a] pyrene-DNA adduct formation in MCF-7 cells exposed to benzo [a] pyrene
Einem Lindeman T, et al.
Mutagenesis, 26(5), 629-635 (2011)
A new selective and potent inhibitor of human cytochrome P450 1B1 and its application to antimutagenesis
Chun YJ, et al.
Cancer Research, 61(22), 8164-8170 (2001)
Maria Tsachaki et al.
Cellular and molecular life sciences : CMLS, 77(6), 1153-1175 (2019-07-16)
Metabolic reprogramming of tumor cells involves upregulation of fatty acid (FA) synthesis to support high bioenergetic demands and membrane synthesis. This has been shown for cytosolic synthesis of FAs with up to 16 carbon atoms. Synthesis of long-chain fatty acids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service