Skip to Content
Merck
All Photos(1)

Documents

SMB00221

Sigma-Aldrich

4,5-Di-O-caffeoylquinic acid

≥85% (LC/MS-ELSD)

Synonym(s):

4,5-DCQA, 4,5-Di-O-caffeoylquinate, 4,5-Dicaffeoylquinic acid, Isochlorogenate C, Isochlorogenic acid C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24O12
CAS Number:
Molecular Weight:
516.45
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥85% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O[C@@H]1C[C@@](O)(C[C@@H](OC(=O)\C=C\c2ccc(O)c(O)c2)[C@H]1OC(=O)\C=C\c3ccc(O)c(O)c3)C(O)=O

InChI

1S/C25H24O12/c26-15-5-1-13(9-17(15)28)3-7-21(31)36-20-12-25(35,24(33)34)11-19(30)23(20)37-22(32)8-4-14-2-6-16(27)18(29)10-14/h1-10,19-20,23,26-30,35H,11-12H2,(H,33,34)/b7-3+,8-4+/t19-,20-,23+,25-/m1/s1

InChI key

UFCLZKMFXSILNL-RVXRWRFUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,5-Di-O-caffeoylquinic acid (4,5-COQ) is a derivative of caffeoylquinic acid which is a bioactive phenylpropanoid.

Application

4,5-Di-O-caffeoylquinic acid has been used as a reference standard to detect phenolic compounds from Artemisia species using high-performance liquid chromatography with diode array detection (HPLC-DAD) It has also been used as a standard in high-performance liquid chromatography with diode array detection (HPLC-DAD) respectively.

Biochem/physiol Actions

4,5-Di-O-caffeoylquinic acid (4,5-COQ) is an excellent antioxidant, anti-inflammatory, antimicrobial, antifungal, and cytoprotective agent. It also shows neuroprotective and anti-amyloidogenic properties and can inhibit 42-mer amyloid β-protein (Aβ42). 4,5-COQ displays anti-human immunodeficiency virus (HIV) activity as its effectively inhibits HIV-1 reverse transcriptase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

HIV-1 reverse transcriptase inhibitory activity of flavones and chlorogenic acid derivatives from Moquiniastrum floribundum (Asteraceae)
Tamayose CI, et al.,
South African Journal of botony, 123, 142-146 (2019)
Phenolic compounds and in vitro antioxidant, anti-inflammatory, antimicrobial activities of Scorzonera hieraciifolia Hayek roots
Sari A, et al.,
Asia-Pacific Journal of Molecular Biology and Biotechnology, 125, 116-119 (2019)
HIV-1 reverse transcriptase inhibitory activity of flavones and chlorogenic acid derivatives from Moquiniastrum floribundum (Asteraceae)
Tamayose CI, et al.,
Asia-Pacific Journal of Molecular Biology and Biotechnology, 123, 142-146 (2019)
Yinku Liang et al.
Molecular medicine reports, 21(1), 229-239 (2019-11-21)
In this study, six compounds were isolated and purified from dandelion, and only sample I exhibited notable antifungal effect on Candida albicans (CA). high‑performance liquid chromatography‑diode‑array detector‑electrospray ionization‑tandem mass spectrometry analysis showed that sample I comprised 4‑coumaric acid, ferulic acid, quercetin
Phenolic compounds and in vitro antioxidant, anti-inflammatory, antimicrobial activities of Scorzonera hieraciifolia Hayek roots
Sari A, et al.,
South African Journal of botony, 125, 116-119 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service