Skip to Content
Merck
All Photos(1)

Key Documents

SMB00200

Sigma-Aldrich

Luteolin 7-rutinoside

≥95% (LC/MS-ELSD)

Synonym(s):

Luteolin-7-O-rhamnoside, Luteoline-7-rhamnoglucoside, Scolymoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O15
CAS Number:
Molecular Weight:
594.52
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](Oc3cc(O)c4C(=O)C=C(Oc4c3)c5ccc(O)c(O)c5)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H30O15/c1-9-20(32)22(34)24(36)26(39-9)38-8-18-21(33)23(35)25(37)27(42-18)40-11-5-14(30)19-15(31)7-16(41-17(19)6-11)10-2-3-12(28)13(29)4-10/h2-7,9,18,20-30,32-37H,8H2,1H3/t9-,18+,20-,21+,22+,23-,24+,25+,26+,27+/m0/s1

InChI key

MGYBYJXAXUBTQF-FOBVWLSUSA-N

General description

Luteolin 7-rutinoside, also known as scolymoside (SCL), is one of the most abundant flavonoids present in an endemic South-African fynbos plant, Cyclopia subternata.

Application

Luteolin 7-rutinoside has been used in reverse phase high performance liquid chromatography with diode array detector (RP-HPLC-DAD) experiment to quantify phenolic compounds.

Biochem/physiol Actions

Luteolin 7-rutinoside has been used in the production of the South African herbal tea honeybush. Aqueous extracts of this flavonoid is used in food industry. Luteolin 7-rutinoside also possess anticoagulant and antiplatelet activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antitcoagulant and antiplatelet activities of scolymoside
Yoon E K, et al.
Bmb Reports, 48(10), 577-577 (2015)
Effect of Low Temperature Cultivation on the Phytochemical Profile and Bioactivity of Arctic Plants: A Case of Dracocephalum palmatum
Olennikov D N, et al.
International Journal of Molecular Sciences, 18(12), 2579-2579 (2017)
Eun-Kyung Yoon et al.
BMB reports, 48(10), 577-582 (2015-04-19)
Cyclopia subternata is a medicinal plant commonly used in traditional medicine to relieve pain. Here, the anticoagulant effects of scolymoside, an active compound in C. subternata, were examined by monitoring activated partial thromboplastin time (aPTT), prothrombin time (PT), and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service