Skip to Content
Merck
All Photos(1)

Key Documents

SMB00149

Sigma-Aldrich

Quercetin 3-glucuronide

≥95% (LC/MS-ELSD)

Synonym(s):

Quercetin 3-glucuronide, Quercetol 3-O-glucuronide, Quercetol glucuronide, Querciturone, 3,3′,4′,5,7-Pentahydroxyflavone 3-glucuronide, Mikwelianin, Miquelianin, Quercetin 3-O-β-D-glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O13
CAS Number:
Molecular Weight:
478.36
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4

InChI

1S/C21H18O13/c22-7-4-10(25)12-11(5-7)32-17(6-1-2-8(23)9(24)3-6)18(13(12)26)33-21-16(29)14(27)15(28)19(34-21)20(30)31/h1-5,14-16,19,21-25,27-29H,(H,30,31)/t14-,15-,16+,19-,21+/m0/s1

InChI key

DUBCCGAQYVUYEU-ZUGPOPFOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quercetin 3-glucuronide is a natural product derived from a plant source. Quercetin is an important flavonoid present in most of the vegetables including onion.[1] It is one of the polyphenol metabolites that is found to be localized to the brain.[2]

Biochem/physiol Actions

Quercetin 3-glucuronide might reduce the risk of developing Alzheimer′s disease dementia. It is found to prevent the formation of β-amyloid (Aβ) peptides in mouse model.[2] Quercetin 3-glucuronide is a metabolic product of dietary quercetin, which occurs in intestinal mucosa and liver. Quercetin 3-glucuronide possess antioxidant activity and is known to have a protective action on LDL (low density lipoprotein) oxidation.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Antioxidant capacity of albumin-bound quercetin metabolites after onion consumption in humans
Murota K, et al.
The Journal of Medical Investigation, 54(3,4) (2007)
Lap Ho et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(2), 769-781 (2012-10-26)
Epidemiological and preclinical studies indicate that polyphenol intake from moderate consumption of red wines may lower the relative risk for developing Alzheimer's disease (AD) dementia. There is limited information regarding the specific biological activities and cellular and molecular mechanisms by

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service