Skip to Content
Merck
All Photos(1)

Key Documents

SMB00100

Sigma-Aldrich

Bavachinin

≥95% (LC/MS-ELSD)

Synonym(s):

4′-Hydroxy-7-methoxy-6-(3-methyl-2-butenyl)flavanone, 7-O-Methylbavachin, Bavachinin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22O4
CAS Number:
Molecular Weight:
338.40
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1cc2O[C@@H](CC(=O)c2cc1C\C=C(\C)C)c3ccc(O)cc3

InChI

1S/C21H22O4/c1-13(2)4-5-15-10-17-18(23)11-20(14-6-8-16(22)9-7-14)25-21(17)12-19(15)24-3/h4,6-10,12,20,22H,5,11H2,1-3H3/t20-/m0/s1

InChI key

VOCGSQHKPZSIKB-FQEVSTJZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Liu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 893-894, 21-28 (2012-03-27)
A fully automated on-line solid-phase extraction (SPE) and high-performance liquid chromatography (HPLC) with diode array detection (DAD) method was developed for determination of bavachinin in mouse plasma. Analytical process was performed on two reversed-phase columns (SPE cartridge and analytical column)
Hiroyuki Haraguchi et al.
Phytotherapy research : PTR, 16(6), 539-544 (2002-09-19)
A meroterpene and four flavonoids were isolated from the seeds of Psoralea corylifolia as antioxidative components. Their structures were elucidated by spectral data and identified as bakuchiol (1), bavachinin (2), bavachin (3), isobavachin (4) and isobavachalcone (5). In particular, meroterpene
Ming Hong Lee et al.
Biological & pharmaceutical bulletin, 28(12), 2253-2257 (2005-12-06)
The overproduction of nitric oxide (NO) by inducible nitric oxide synthase (iNOS) switches the function of NO from a physiological neuromodulator to a neurotoxic effector in central nervous system (CNS) after brain injury. From the methanol extracts of Psoralea corylifolia
Xiumin Chen et al.
FEBS letters, 587(18), 2930-2935 (2013-08-03)
Spontaneous aggregation of Aβ is a key factor in the development of Alzheimer's disease. In searching for Aβ aggregation inhibitors from traditional Chinese herbal medicines, we identified two active compounds from Psoraleae Fructus, namely isobavachalcone and bavachinin. We further demonstrated
Jianmei Luo et al.
Journal of bioscience and bioengineering, 117(2), 191-196 (2013-09-10)
Biotransformation of bavachinin (1) was investigated using three fungal cell cultures of Aspergillus flavus ATCC 30899, Cunninghamella elegans CICC 40250 and Penicillium raistrickii ATCC 10490, respectively. Two major converted products were identified by LC/MS, (1)H NMR and (13)C NMR and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service