Skip to Content

SD6A

Latex beads, styrene divinylbenzene

aqueous suspension, 6.4 μm mean particle size

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41100000
NACRES:
NB.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

aqueous suspension

composition

Solids, 10%

mean particle size

6.4 μm



Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library