Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700447

Sigma-Aldrich

Monoclonal Anti-MYC , (C-terminal) antibody produced in mouse

clone 9E10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Cellular myelocytomatosis oncogene, Anti-c-Myc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9E10, monoclonal

form

buffered aqueous solution

species reactivity

fusion proteins in all species, human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

General description

The cellular myelocytomatosis (c-myc) gene mapped to human chromosome 8q24, is the cellular homologue of the v-myc gene originally isolated from an avian myelocytomatosis virus. c-myc is a member of MYC gene family. c-Myc gene codes for basic helix-loop-helix/leucine zipper (bHLH/LZ) transcription factor that regulates the G1-S cell cycle transition.

Immunogen

Synthetic peptide sequence (AEEQKLISEEDLL) corresponding to the C-terminal region of human c-Myc

Application

Monoclonal Anti-MYC, (C-terminal) antibody produced in mouse has been used in following studies:
  • western blotting analysis
  • chromatin immunoprecipitation quantitative PCR (ChIP-qPCR assays
  • immunofluorescence microscopy
The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 1-5 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator. Membrane permeabilization is required.

Biochem/physiol Actions

The cellular myelocytomatosis (c-myc) oncogene plays a vital role in cellular proliferation, differentiation, apoptosis and acts as transcriptional regulator of gene expression. c-Myc expression is essential and sufficient to assist most of the cells to enter DNA synthetic (S) phase of the cell cycle. The encoded protein plays a crucial role in vasculogenesis and angiogenesis during cancer development and progression. c-Myc interacts with its binding partner Max and activates the transcription of growth promoting genes such as cyclin D2, ornithine decarboxylase and E2F1 and it also represses the transcription of multiple genes, especially p21 and p27, by binding to the transcription initiator element (Inr) in a complex with Max and either Sp1 or Miz1. Overexpression of MYC in DLBCL (diffuse large B-cell lymphoma) results in poor outcome and invasive treatment when medicated with rituximab plus cyclophosphamide, doxorubicin, vincristine and prednisone (R-CHOP).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CSBF/C10orf99, a novel potential cytokine, inhibits colon cancer cell growth through inducing G1 arrest
Pan W
Scientific Reports (2014)
Irene Zornetta et al.
Toxins, 9(3) (2017-03-08)
Scorpions are among the oldest venomous living organisms and the family Buthidae is the largest and most medically relevant one. Scorpion venoms include many toxic peptides, but recently, a metalloprotease from Tityus serrulatus called antarease was reported to be capable
Apoptotic signaling by c-MYC.
Hoffman B and Liebermann DA.
Oncogene, 27, 6462-6472 (2008)
Interferon-Inducible Oligoadenylate Synthetase-Like Protein Acts as an Antiviral Effector against Classical Swine Fever Virus via the MDA5-Mediated Type I Interferon-Signaling Pathway
Li LF
Journal of Virology, 91, e01514-e01516 (2017)
Positive feedback promotes mitotic exit via the APC/C-Cdh1-separase-Cdc14 axis in budding yeast
Hatano Y
Cellular Signalling, 28, 1545-1554 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service