Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503373

Sigma-Aldrich

Anti-TEAD2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

TEA domain family member 2, TEAD-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 49 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TEAD2(8463)

Related Categories

General description

TEA domain transcription factor 2 (TEAD2) belongs to the TEAD family. TEAD2 gene is located on human chromosome 19q13.3 Anti-TEAD2 antibody detects endogenous levels of total TEAD2 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human TEAD2.

Immunogen Range: 71-120

Application

Anti-TEAD2 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

TEA domain transcription factor 2 (TEAD2) plays a regulatory role in cell stemness. It modulates the expression of several genes. TEAD2 participates in the maintenance of cell survival. It is essential during neural development. TEAD2 might be a new therapeutic target and a predictive indicator of hepatocellular carcinoma (HCC).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kotaro J Kaneko et al.
Genesis (New York, N.Y. : 2000), 45(9), 577-587 (2007-09-18)
TEAD2, one of the first transcription factors expressed at the beginning of mammalian development, appears to be required during neural development. For example, Tead2 expression is greatest in the dorsal neural crest where it appears to regulate expression of Pax3
Yanli Wang et al.
Molecular medicine reports, 20(4), 3519-3526 (2019-09-06)
Cisplatin has been widely used as a conventional treatment for patients with non‑small cell lung cancer (NSCLC). However, primary and acquired cisplatin resistances are frequently developed during the treatment of patients with NSCLC, leading to an increased mortality rate. Accumulating
Jong Seok Joo et al.
Oncology reports, 43(6), 1785-1796 (2020-04-24)
TEA Domain Transcription Factors (TEADs) are important in development and serve essential roles in tumorigenesis; however, the role of TEAD2 expression in hepatocellular carcinoma (HCC) has not been widely examined. The present study was conducted to investigate the expression status
Localization of human transcription factor TEF-4 and TEF-5 (TEAD2, TEAD3) genes to chromosomes 19q13.3 and 6p21.2 using fluorescence in situ hybridization and radiation hybrid analysis.
P Jacquemin et al.
Genomics, 55(1), 127-129 (1999-01-16)
Paula González-Alonso et al.
Cancers, 12(5) (2020-05-06)
Trastuzumab is the first-line targeted therapeutic drug for HER2-positive breast cancer, leading to improved overall survival. However, acquired resistance inevitably occurs. We aimed to identify, quantify, and assess the mechanisms of acquired resistance to trastuzumab. We established an acquired trastuzumab-resistant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service