Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502989

Sigma-Aldrich

Anti-TNF Receptor II, antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CD120b, p75, p75 TNF receptor, p75TNFR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNFRSF1B(7133)

General description

Tumor necrosis factor receptor 2 (TNFR2) gene with ten exons and nine introns, spanning approximately 43kbp is mapped to human chromosome 1p36.3-p36.2. TNFR2 is a member of TNF receptor superfamily, and it is expressed highly on cells of hematopoietic lineage. TNFR2, also known as CD120b or p75, TNF receptor is a 75-kDa type I transmembrane protein, characterized with cysteine-rich extracellular regions with three to six disulfide-linked domains required for ligand binding.

Immunogen

The antiserum was produced against synthesized peptide derived from human TNF Receptor II.

Immunogen Range: 376-425

Application

Anti-TNF Receptor 2 antibody has been used to study the effect of TNF signalling on CPF- and CPF+ CM (pesticides)-induced toxicity or apoptosis in SH-SY5Y cells.

Biochem/physiol Actions

Tumor necrosis factor receptor 2 (TNFR2) plays a vital role in inducing signals for various cellular process, including proliferation of thymocytes and peripheral T cells, T cell secretion of granulocyte-macrophage colony-stimulating factor, inhibition of early hematopoiesis and the inactivation and clearance of TNF by the kidney. Mutation in the gene leads to hematopoietic cell malignancies, neuroblastoma, glioma and cervical and ovarian carcinoma. TRAF2 interacts with TNFR2 and activates NF-κB (NF-κB) pathway. TNFR2, which lack death domains modulates the effect of other death receptor TNFR1, which induces T cell death. TNFR2 plays an essential role in TNFα–induced leukocyte-endothelial interaction. TNFR2 is considered as a potential therapeutic alternative, as it has capacity to nullify the common side effects associated with drugs targeting TNF-α or p55.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Jiang et al.
Experimental and therapeutic medicine, 15(1), 1021-1028 (2018-02-13)
Tumor necrosis factor (TNF)-α has been reported to be important in glomerulonephritis, which is closely associated with podocyte dysfunction and apoptosis. However, the precise mechanisms by which TNF-α expression are regulated remain unclear. The purpose of the present study was
S M Santee et al.
The Journal of biological chemistry, 271(35), 21151-21159 (1996-08-30)
Tumor necrosis factor receptor p75 (TNF-R p75) is a 75-kDa type I transmembrane protein expressed predominantly on cells of hematopoietic lineage. TNF-R p75 belongs to the TNF receptor superfamily characterized by cysteine-rich extracellular regions composed of three to six disulfide-linked
Unni M Chandrasekharan et al.
Blood, 109(5), 1938-1944 (2006-10-28)
Tumor necrosis factor-alpha (TNF-alpha) binds to 2 distinct cell-surface receptors: TNF-alpha receptor-I (TNFR-I: p55) and TNF-alpha receptor-II (TNFR-II: p75). TNF-alpha induces leukocyte adhesion molecules on endothelial cells (ECs), which mediate 3 defined steps of the inflammatory response; namely, leukocyte rolling
Bence Pál Barta et al.
World journal of diabetes, 14(1), 48-61 (2023-01-24)
Cytokines are essential in autoimmune inflammatory processes that accompany type 1 diabetes. Tumor necrosis factor alpha plays a key role among others in modulating enteric neuroinflammation, however, it has a dual role in cell degeneration or survival depending on different
M Rothe et al.
Science (New York, N.Y.), 269(5229), 1424-1427 (1995-09-08)
TNF receptor-associated factor (TRAF) proteins are candidate signal transducers that associate with the cytoplasmic domains of members of the tumor necrosis factor (TNF) receptor superfamily. The role of TRAFs in the TNF-R2 and CD40 signal transduction pathways, which result in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service