Skip to Content
Merck
All Photos(3)

Key Documents

SAB4502966

Sigma-Aldrich

Anti-Tyrosine Hydroxylase, N-Terminal antibody produced in rabbit

affinity isolated antibody

Synonym(s):

TH isoform 3, TH isoform a, TH-4, TY3H, TYH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 58 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TH(7054)

Related Categories

General description

Anti-Tyrosine Hydroxylase Antibody detects endogenous levels of total Tyrosine Hydroxylase protein.
The TH (tyrosine Hydroxylase) gene codes for a homotetramer that contains catalytic, regulatory, and tetramerization domains in each of its subunit.

Immunogen

The antiserum was produced against synthesized peptide derived from human Tyrosine Hydroxylase.

Immunogen Range: 41-90

Application

Anti-Tyrosine Hydroxylase, N-Terminal antibody produced in rabbit has ben used in immunohistochemistry.

Biochem/physiol Actions

The conversion of l-tyrosine into l-DOPA (L-3,4-dihydroxyphenylalanine) is catalyzed by tyrosine hydroxylase (TH), which is the rate-limiting step in the in the synthesis of catecholamines such as dopamine and also other necessary hormones and neuromodulators. The TH enzyme deficiency results in low levels of dopamine, which is characterised in Parkinson′s disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrenergic regulation during acute hepatic infection with Entamoeba histolytica in the hamster: involvement of oxidative stress, Nrf2 and NF-KappaB.
Aldaba-Muruato LR
Parasite (Paris, France), 24:46, 1-13 (2017)
Regulation of tyrosine hydroxylase is preserved across different homo- and heterodimeric 14-3-3 proteins.
Ghorbani S
Amino Acids, 48(5), 1221-1229 (2016)
Molecular Mechanism of Regulation of MTA1 Expression by Granulocyte Colony-stimulating Factor.
Kumar AS
The Journal of Biological Chemistry, 291(23), 12310-12321 (2016)
Phosphorylation at serine 31 targets tyrosine hydroxylase to vesicles for transport along microtubules.
Jorge-Finnigan A
The Journal of Biological Chemistry, 292(34), 14092-14107 (2017)
Izel Tekin et al.
Parkinsonism & related disorders, 23, 86-90 (2016-01-07)
Many of the symptoms and signs of Parkinson's disease (PD) arise from the death of midbrain dopamine neurons that utilize tyrosine hydroxylase (TH) as the rate-limiting enzyme in catecholamine biosynthesis. We investigated whether the presence of a common TH polymorphism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service