Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502910

Sigma-Aldrich

Anti-TACC1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

KIAA1103, transforming acidic coiled-coil-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 87 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TACC1(6867)

General description

Anti-TACC1 Antibody detects endogenous levels of total TACC1 protein.
The gene TACC1 (transforming acidic coiled-coil containing protein 1) is mapped to human chromosome 8p11. It is highly expressed during embryogenesis and is weakly expressed in differentiated tissues. TACC1 belongs to the TACC family of proteins and localizes in the nucleus.

Immunogen

The antiserum was produced against synthesized peptide derived from human TACC1.

Immunogen Range: 11-60

Biochem/physiol Actions

TACC1 (transforming acidic coiled-coil containing protein 1) participates in the transcriptional regulation of nuclear hormone receptors (NRs). It is associated with breast and ovarian cancers and leukemia.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan J Wilde et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(10), 2565-2579 (2017-02-06)
Diencephalic defects underlie an array of neurological diseases. Previous studies have suggested that retinoic acid (RA) signaling is involved in diencephalic development at late stages of embryonic development, but its roles and mechanisms of action during early neural development are
Romain Guyot et al.
BMC molecular biology, 11, 3-3 (2010-01-19)
The transcriptional activity of Nuclear hormone Receptors (NRs) is regulated by interaction with coactivator or corepressor proteins. Many of these cofactors have been shown to have a misregulated expression or to show a subcellular mislocalization in cancer cell lines or
Omkaram Gangisetty et al.
Oncogene, 23(14), 2559-2563 (2004-02-10)
Dysregulation of the human transforming acidic coiled coil (TACC) genes is thought to be important in the development of multiple myeloma, breast and gastric cancer. However, even though these proteins have been implicated in the control of cell growth and
Aiping Ding et al.
PloS one, 8(12), e82136-e82136 (2013-12-21)
NPM, TFF3 and TACC1 are molecular markers that play important roles in cell differentiation. Herein, we investigated their prognostic impact in patients with primary gastric cancer (GC) and determined whether they could be used as markers of more aggressive gastric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service