Skip to Content
Merck
All Photos(3)

Key Documents

SAB4502676

Sigma-Aldrich

Anti-S6 Ribosomal Protein, C-Terminal antibody produced in rabbit

affinity isolated antibody

Synonym(s):

40S ribosomal protein S6, Phosphoprotein NP33, RPS6, RS6, ribosomal protein S6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 28 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPS6(6194)

General description

Anti-S6 Ribosomal protein antibody detects endogenous levels of total S6 Ribosomal protein protein.
The RPS6 (ribosomal protein S6) gene is mapped to human chromosome 9p22.1.

Immunogen

The antiserum was produced against synthesized peptide derived from human S6 Ribosomal Protein.

Immunogen Range: 191-240

Biochem/physiol Actions

The RPS6 (ribosomal protein S6) protein is involved in the 40S subunit assembling. RpS6 might be involved in the regulation of cellular metabolism, the size of cells, their proliferation and survival. RpS6 might stimulate or inhibit translation of certain mRNA. Upregulation of the gene is observed in pulmonary sclerosing hemangioma, lymphangioleiomyomatosis-associated angiomyolipomas, dysplasia, oral cavity and esophageal squamous cell carcinoma.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janice Chithelen et al.
Frontiers in physiology, 13, 856143-856143 (2022-04-05)
The bioactive sphingolipids ceramide and sphingosine-1-phosphate (S1P) are involved in the regulation of cell homeostasis and activity ranging from apoptosis to proliferation. We recently described that the two compounds ceranib-2 (inhibiting acid ceramidase) and SKI-II [inhibiting the sphingosine kinases 1
Hyperphosphorylation of ribosomal protein S6 predicts unfavorable clinical survival in non-small cell lung cancer.
Chen B
Journal of Experimental & Clinical Cancer Research, 34:126, 1-16 (2015)
Aggregation of Ribosomal Protein S6 at Nucleolus Is Cell Cycle-Controlled and Its Function in Pre-rRNA Processing Is Phosphorylation Dependent.
Zhang D
Journal of Cellular Biochemistry, 117(7), 1649-1657 (2016)
Potential therapeutic targets for chordoma: PI3K/AKT/TSC1/TSC2/mTOR pathway.
Presneau N
British Journal of Cancer, 100(9), 1406-1414 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service