Skip to Content
Merck
All Photos(3)

Key Documents

SAB4502491

Sigma-Aldrich

Anti-COX2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

COX2, CYCLOOXYGENASE 2, GLUCOCORTICOID-REGULATED INFLAMMATORY PROSTAGLANDIN G/H SYNTHASE, GRIPGHS, PGHS2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 68 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTGS2(5743)

Related Categories

General description

Anti-COX2 Antibody detects endogenous levels of total COX2 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Cox2.

Immunogen Range: 555-604

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadine Ansorge et al.
Diagnostics (Basel, Switzerland), 11(3) (2021-04-04)
Vulvar cancer incidence numbers have been rising steadily over the past decades. Especially the number of young patients with vulvar cancer increased recently. Therefore, the need to identify new prognostic factors for vulvar carcinoma is more apparent. Cyclooxygenase-2 (COX-2) has
Nadine Ansorge et al.
International journal of molecular sciences, 23(9) (2022-05-15)
Vulvar cancer incidence numbers have been steadily rising over the past decades. In particular, the number of young patients with vulvar cancer has recently increased. Therefore, the need to identify new prognostic factors and, in addition, therapeutic options for vulvar
Wanting Shao et al.
Journal of translational medicine, 18(1), 94-94 (2020-02-23)
The aim of this study was to investigate the expression of the nuclear receptor PPARγ, together with that of the cyclooxygenases Cox-1 and Cox-2, in breast cancer (BC) tissues and to correlate the data with several clinicobiological parameters including patient
Yao Ye et al.
Endocrine connections, 7(5), 749-761 (2018-04-28)
Although an inflammatory microenvironment is required for successful implantation, an inflammatory overreaction is one of the causes of unexplained recurrent pregnancy losses (uRPL). Prostaglandin E2 (PGE2) plays a pivotal role in regulating immune balance during early pregnancy, and it can
Ilona Kowalczyk-Zieba et al.
Biology of reproduction, 90(1), 17-17 (2013-12-07)
Lysophosphatidic acid (LPA) together with its active G protein-coupled receptors are present in the corpus luteum (CL) of the cow. Under in vivo conditions, LPA stimulated P4 and PGE2 secretion during the luteal phase of the estrous cycle in heifers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service