Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502211

Sigma-Aldrich

Anti-PLK1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

PLK-1, Polo-like kinase 1, STPK13, Serine/threonine-protein kinase 13

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 68 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLK1(5347)

Related Categories

General description

Anti-PLK1 Antibody detects endogenous levels of total PLK1 protein.
PLK1 (polo like kinase 1) gene is mapped to human chromosome 16p12.2. It codes for the protein serine/threonine kinase.

Immunogen

The antiserum was produced against synthesized peptide derived from human PLK1.

Immunogen Range: 176-225

Biochem/physiol Actions

PLK1 (polo like kinase 1) is predominantly involved in mitosis. It mediates the phosphorylation of cyclin B1/CDK1 (cyclin dependent kinase 1) complex, and activation of the anaphase promoting complex. PLK1 has anti-apoptotic function. Upregulation of gene is observed in different types of cancer, such as colorectal cancer and is regarded as an oncogene. PLK1 is involved in checkpoint recovery and adaptation following DNA damage.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of the microtubule-associated protein MAP9/ASAP and its partners AURKA and PLK1 in colorectal and breast cancers.
Rouquier S
Disease Markers, 2014:798170, 1-6 (2014)
Polo-like kinase 1 expression is suppressed by CCAAT/enhancer-binding protein ? to mediate colon carcinoma cell differentiation and apoptosis.
Dasgupta N
Biochimica et Biophysica Acta, 1861(7), 1777-1787 (2017)
Polo-like Kinase 1 as a potential therapeutic target in Diffuse Intrinsic Pontine Glioma.
Amani V
BMC Cancer, 16:647, 1-9 (2016)
PLK1 inhibition enhances temozolomide efficacy in IDH1 mutant gliomas.
Koncar RF
Oncotarget, 8(9), 15827-15837 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service