Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502050

Sigma-Aldrich

Anti-OR1D2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

OR17-4, Olfactory receptor 17-4, Olfactory receptor 1D2, Olfactory receptor OR17-6, Olfactory receptor-like protein HGMP07E

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 35 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OR1D2(4991)

General description

Anti-OR1D2 antibody detects endogenous levels of total OR1D2 protein.
The OR1D2 (olfactory receptor family 1 subfamily D member 2) gene is located on human chromosome 17p13.3. The gene is considered as the first olfactory receptor expressed in the nose. It is also found to be expressed in spermatozoa. The encoded protein is a member of G protein-coupled receptor superfamily and exists as a homo-oligomer.

Immunogen

The antiserum was produced against synthesized peptide derived from human OR1D2.

Immunogen Range: 201-250

Biochem/physiol Actions

The protein encoded by OR1D2 (olfactory receptor family 1 subfamily D member 2) gene takes part in the process of fertilization by mediating the communication between the sperm and egg via chemotaxis. Sperm expressing OR1D2 is known to migrate toward odorant ligands such as lilial, floralozone and bourgeonal.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Ottaviano et al.
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery, 270(12), 3079-3086 (2013-03-26)
Olfactory receptor (OR) expression is also present in the sperm cells and could mediate sperm chemotaxis. OR1D2 was the first OR expressed in the testis demonstrated to be involved in chemotaxis and to be expressed also in the nose with
Brian L Cook et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(29), 11925-11930 (2009-07-08)
Although understanding of the olfactory system has progressed at the level of downstream receptor signaling and the wiring of olfactory neurons, the system remains poorly understood at the molecular level of the receptors and their interaction with and recognition of
Marc Spehr et al.
Science (New York, N.Y.), 299(5615), 2054-2058 (2003-03-29)
Although it has been known for some time that olfactory receptors (ORs) reside in spermatozoa, the function of these ORs is unknown. Here, we identified, cloned, and functionally expressed a previously undescribed human testicular OR, hOR17-4. With the use of
Fallou Wade et al.
The Journal of biological chemistry, 286(17), 15252-15259 (2011-04-02)
G-protein-coupled receptor homo-oligomerization has been increasingly reported. However, little is known regarding the relationship between activation of the receptor and its association/conformational states. The mammalian olfactory receptors (ORs) belong to the G protein-coupled receptor superfamily. In this study, the homo-oligomerization
A partial deletion of the aspartoacylase gene is the cause of Canavan disease in a family from Mexico.
F E Tahmaz et al.
Journal of medical genetics, 38(3), E9-E9 (2001-03-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service