Skip to Content
Merck
All Photos(1)

Documents

SAB4501582

Sigma-Aldrich

Anti-Integrin β1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CD29, FNRB, Fibronectin receptor β subunit, ITB1, Integrin VLA-4 β subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 88 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)

Related Categories

General description

Anti-Integrin β1 Antibody detects endogenous levels of total Integrin β1 protein.
Integrinβ1 is a mechanosensor, that is located on human chromosome 10p11.22. It is present in normal cells and tumor-associated cells.

Immunogen

The antiserum was produced against synthesized peptide derived from human Integrin beta1.

Immunogen Range: 749-798

Application

Anti-integrin β1, C-terminal antibody has been used in immunophenotyping of separated cells.

Biochem/physiol Actions

Integrin β1 acts as an adhesive molecule in the development of de novo focal contact, keratinocyte migration and re-epithelialization of wound tissue.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of mesenchymal stem cells in treatment of infertility in male rats.
Hassan AI and Alam SS
Stem Cell Research & Therapy, 5(6), 131-131 (2014)
β1 Integrin as a Prognostic and Predictive Marker in Triple-Negative Breast Cancer.
Yin HL, et al.
International Journal of Molecular Sciences, 17(9), 1432-1432 (2016)
β1 integrin signaling in asymmetric migration of keratinocytes under mechanical stretch in a co-cultured wound repair model.
Lu D, et al.
BioMedical Engineering OnLine, 15(2), 130-130 (2016)
The Effect of Mesenchymal Stem Cells and Chitosan Gel on Full Thickness Skin Wound Healing in Albino Rats: Histological, Immunohistochemical and Fluorescent Study.
El Sadik AO, et al.
PLoS ONE, 10(9) (2015)
Genomic aberrations associated with erlotinib resistance in non-small cell lung cancer cells.
Serizawa M, et al.
Anticancer Research, 33(12), 5223-5233 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service