Skip to Content
Merck
All Photos(1)

Key Documents

SAB4501408

Sigma-Aldrich

Anti-HNF4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

HNF-4, HNF-4-α, HNF4, HNF4A, Hepatocyte nuclear factor 4-α

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 52 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HNF4A(3172)

General description

Anti-HNF4 Antibody detects endogenous levels of total HNF4 protein.
Hepatocyte nuclear factor 4 alpha (HNF4α) is a transcription factor, located on human chromosome 20q13.12.

Immunogen

The antiserum was produced against synthesized peptide derived from human HNF4 alpha.

Immunogen Range: 280-329

Biochem/physiol Actions

Hepatocyte nuclear factor 4 alpha (HNF4α) plays an important role in the growth of liver. Mutations in HNF4α results in maturity onset diabetes of the young (MODY).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association study of genetic polymorphisms of SLC2A10 gene and type 2 diabetes in the Taiwanese population.
Lin WH, et al.
Diabetologia, 49(6), 1214-1221 (2006)
The Common p.R114W HNF4A Mutation Causes a Distinct Clinical Subtype of Monogenic Diabetes.
Laver TW, et al.
Diabetes, 65(10), 3212-3217 (2016)
Direct induction of hepatocyte-like cells from immortalized human bone marrow mesenchymal stem cells by overexpression of HNF4α.
Hu X, et al.
Biochemical and Biophysical Research Communications, 478(2), 791-797 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service