Skip to Content
Merck
All Photos(3)

Documents

SAB4501175

Sigma-Aldrich

Anti-Connexin 43 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CX43, CXA1, CXN-43, Connexin 43, Gap junction 43 kDa heart protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 43 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GJA1(2697)

Related Categories

General description

Anti-Connexin 43 Antibody detects endogenous levels of total Connexin 43 protein.
Connexin 43 (Cx43) is also known as GJA1 (gap junction protein α 1) and belongs to a large family of homologous connexin proteins. It has an intracellular N-terminus, four transmembrane domains, two extracellular loops, one cytoplasmic loop and an intracellular C-terminus. It is located on human chromosome 6q22-q23.

Immunogen

The antiserum was produced against synthesized peptide derived from human Connexin 43.

Immunogen Range: 333-382

Biochem/physiol Actions

Mutations in Connexin 43 (Cx43) gene result in pleiotropic phenotype of oculodentodigital dysplasia. Connexin 43 participates in the multiplication and differentiation of mature T cells. It also participates in the secretion and manufacturing of cytokines. It helps to release lactate from glycolytic pancreatic ductal adenocarcinoma cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Up-regulation of gap junction in peripheral blood T lymphocytes contributes to the inflammatory response in essential hypertension
Ni X, et al.
PLoS ONE, 12 (2017)
Connexin 43 (GJA1) mutations cause the pleiotropic phenotype of oculodentodigital dysplasia
Paznekas WA, et al.
American Journal of Human Genetics, 72, 408-418 (2003)
Camilla Schinner et al.
JCI insight, 5(9) (2020-05-08)
Arrhythmogenic cardiomyopathy (AC) is a genetic disease causing arrhythmia and sudden cardiac death with only symptomatic therapy available at present. Mutations of desmosomal proteins, including desmoglein-2 (Dsg2) and plakoglobin (Pg), are the major cause of AC and have been shown
Connexin-43 channels are a pathway for discharging lactate from glycolytic pancreatic ductal adenocarcinoma cells
Dovmark TH, et al.
Oncogene, 36, 4538-4550 (2017)
Mohammad El-Harakeh et al.
World journal of gastroenterology, 28(40), 5845-5864 (2022-11-11)
Inflammatory bowel disease (IBD) constitutes a substantial risk factor for colorectal cancer. Connexin 43 (Cx43) is a protein that forms gap junction (GJ) complexes involved in intercellular communication, and its expression is altered under pathological conditions, such as IBD and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service