Skip to Content
Merck
All Photos(1)

Key Documents

SAB4301578

Sigma-Aldrich

Anti-phospho-ERK1/2 (pThr202/Tyr204) antibody produced in rabbit

Anti-phospho-ERK1/2 (pThr202/Tyr204) antibody produced in rabbit
1 of 1 reviewers received a sample product or took part in a promotion

affinity isolated antibody

Synonym(s):

ERK-1, ERK1, ERT2, P44ERK1, PRKM3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa
44 kDa

species reactivity

rat, mouse, human

concentration

1.0 mg/mL

technique(s)

western blot: 1:500-1:1000 (Cell Lysate)

isotype

IgG

accession no.

NP_001035145.1 

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr202/pTyr204)

Gene Information

human ... MAPK3(5595)

Specificity

The antibody detects endogenous level of p44/42 MAP Kinase only when phosphorylated at Threonine 202 and Tyrosine 204 .

Immunogen

Peptide sequence around phosphorylation site of Threonine 202/Tyrosine 204(F-L-T(p)-E-Y(p)-V-A) derived from Human p44/42 MAP Kinase.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 1.0mg/mL in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lenka Halova et al.
Open biology, 11(4), 200405-200405 (2021-04-08)
Fluctuations in TOR, AMPK and MAP-kinase signalling maintain cellular homeostasis and coordinate growth and division with environmental context. We have applied quantitative, SILAC mass spectrometry to map TOR and nutrient-controlled signalling in the fission yeast Schizosaccharomyces pombe. Phosphorylation levels at
Varun Kumar et al.
Current eye research, 46(5), 710-718 (2020-10-28)
Understanding molecular changes is essential for designing effective treatments for nonarteritic anterior ischemic optic neuropathy (AION), the most common acute optic neuropathy in adults older than 50 years. We investigated changes in the mitogen-activated protein kinase (MAPK) pathway after experimental AION
Vittorio Perfetti et al.
Oncotarget, 8(34), 56158-56167 (2017-09-17)
Gastrointestinal stromal tumors (GISTs) are the most common mesenchymal tumors of the gastrointestinal tract. GISTs express the receptor tyrosine kinase KIT, and the majority of GISTs present KIT gain-of-function mutations that cluster in the 5' end of the receptor juxtamembrane
Haiying Peng et al.
Frontiers in immunology, 14, 1167605-1167605 (2023-11-29)
TGF-β1 can induce epithelial-mesenchymal transition (EMT) in primary airway epithelial cells (AECs). Semaphorin7A (Sema7a) plays a crucial role in regulating immune responses and initiating and maintaining transforming growth factor β1 TGF-β1-induced fibrosis. To determine the expression of Sema7a, in serum
Li Cao et al.
Microbial pathogenesis, 131, 120-127 (2019-03-27)
Enterovirus 71 (EV71) is the main causative agent of hand, foot, and mouth disease (HFMD), which has high morbidity and mortality. It mainly threatens children under six years of age. Because of a poor understanding of its pathogenesis, there are

Reviews

Active Filters

  1. New Jersey
    • Review 1
    • Votes 0
    1 out of 5 stars.

    Beware of nice picture from the supplier

    The picture you show was the reason I ordered. However the blot looks awful many, many bands and recognize the markers. Very disapointed

    Helpful?

    1. Response from Merck KGaA:

      Thank you for taking the time to leave a review! We are sorry to hear that your experience with this antibody did not meet your expectations. If you have a moment, please contact us by visiting https://www.sigmaaldrich.com/support/customer-support and submit a Product Technical Inquiry so that we can assist you. We look forward to hearing from you soon!

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service