Skip to Content
Merck
All Photos(4)

Documents

SAB4200310

Sigma-Aldrich

Anti-Granulin (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Acrogranin, Anti-Epithelin, Anti-GEP, Anti-GP88, Anti-GRN, Anti-PCDGF, Anti-PEPI, Anti-PGRN, Anti-Progranulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~80 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 10-20 μg/mL using A431 cells
western blot: 1-2 μg/mL using MCF7 cell extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRN(2896)

General description

Granulin is composed of 7.5 repeat units of a unique, structurally defined cysteine-rich granulin-eptihelin motif.
Granulins (also known as granulin/epithelin precursor [GEP]) are a secreted growth factors expressed mainly by granulocytes. They are secreted as high molecular weight precursor proteins, pro-granulins (PGRN), with seven and a half repeats of 12-cysteine granulin motif. Five human granulins have been isolated and they have a common precursor. PGRN is highly expressed in rapidly proliferating epithelial cells such as keratinocytes. It is also synthesised in neurons of cerebral cortex, hippocampus and cerebellum, central and peripheral nervous system.

Immunogen

synthetic peptide corresponding to a sequence at the C-terminus of human granulin precursor, conjugated to KLH.

Application

Anti-Granulin (C-terminal) antibody produced in rabbit has been used in immunoblotting, immunofluorescence and western blotting.

Biochem/physiol Actions

Granulins are potent mitogenic factors, and mediate proliferation, motility, wound healing and anchorage-dependent growth of fibroblasts. Some of the pathways that are associated with PGRN are MAPK and PI3K/Akt.
Progranulin gene is mitotically quiescent epithelia and is expressed in low levels. It stimulates the proliferation in epithelial cells and promotes their anchorage-independent growth. Overexpression of progranulin confers epithelial invasiveness and tumorigenicity. There is growing evidence that granulin is involved in embryonic and neonatal development, placenta, the epidermis, vasculature angiogenesis, and the developing nervous system. Granulin stimulates an increase in cyclins D1 and cyclins B. Granulin overexpression has been associated with many cancers, including glioblastoma, breast, renal and invasive ovarian cancers. Granulin is highly expressed in motor neurons, it regulates neurite outgrowth and promotes neuronal cell survival. Progranulin gene mutations and PGRN gene haploinsufficiency have been recently proposed to cause a form of frontotemporal dementia (FTLD). It is also associated with the formation of pathogenic Tar-DNA Binding Protein 43 (TDP-43) and ubiquitin positive inclusions.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Zanocco-Marani et al.
Cancer research, 59(20), 5331-5340 (1999-10-28)
Growth-regulated cells, such as 3T3 mouse embryo fibroblasts (MEFs), require more than one growth factor for growth, usually the insulin-like growth factor I (IGF-I) in combination with either platelet-derived growth factor or epidermal growth factor. Singly, these growth factors cannot
Progranulin (acrogranin/PC cell-derived growth factor/granulin-epithelin precursor) is expressed in the placenta, epidermis, microvasculature, and brain during murine development
Daniel R, et al.
Developmental Dynamics, 227(4), 593-599 (2003)
Progranulin is expressed within motor neurons and promotes neuronal cell survival
Ryan CL, et al.
BMC Neuroscience, 10(1), 130-130 (2009)
Cellular localization of gene expression for progranulin
Daniel R, et al.
The Journal of Histochemistry and Cytochemistry, 48(7), 999-1009 (2000)
Progranulin-mediated deficiency of cathepsin D results in FTD and NCL-like phenotypes in neurons derived from FTD patients
Valdez C, et al.
Human Molecular Genetics, 26(24), 4861-4872 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service